Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21102298-21102565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM253
Entrez Gene ID
643382
Gene Name
transmembrane protein 253
Gene Aliases
C14orf176, C14orf95, NCRNA00220
UniGene
Hs.540107
Gene Chromosome Location
Chr.14: 21098937-21103724 on Build GRCh38
Transcripts
NM_001146683.1 XM_006720233.3 XM_011537078.2 XM_011537079.1 XM_011537081.2 XM_011537082.2 XM_011537083.2 XM_011537084.2 XM_011537085.2 NM_001102454.1 XM_006720164.3

Gene Symbol
ZNF219
Entrez Gene ID
51222
Gene Name
zinc finger protein 219
Gene Aliases
ZFP219
UniGene
Hs.250493
Gene Chromosome Location
Chr.14: 21090046-21104704 on Build GRCh38
Transcripts
NM_001146683.1 XM_006720233.3 XM_011537078.2 XM_011537079.1 XM_011537081.2 XM_011537082.2 XM_011537083.2 XM_011537084.2 XM_011537085.2 NM_001102454.1 XM_006720164.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5798110 c.331G>T p.V111L Chr.14: 21102459-21102459 on Build GRCh38
1607467 c.287T>G p.M96R Chr.14: 21102415-21102415 on Build GRCh38
5605576 c.355G>A p.A119T Chr.14: 21102483-21102483 on Build GRCh38
1152706 c.364C>T p.P122S Chr.14: 21102492-21102492 on Build GRCh38
5569165 c.373A>G p.T125A Chr.14: 21102501-21102501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563057678 Chr.14: 21102536-21102536 on Build GRCh38
rs773980269 Chr.14: 21102414-21102414 on Build GRCh38
rs768832501 Chr.14: 21102542-21102542 on Build GRCh38
rs748483408 Chr.14: 21102409-21102409 on Build GRCh38
rs768998973 Chr.14: 21102406-21102406 on Build GRCh38
rs769855689 Chr.14: 21102338-21102338 on Build GRCh38
rs979435616 Chr.14: 21102387-21102387 on Build GRCh38
rs932954064 Chr.14: 21102526-21102526 on Build GRCh38
rs989073583 Chr.14: 21102432-21102432 on Build GRCh38
rs949558369 Chr.14: 21102463-21102463 on Build GRCh38
rs4982408 Chr.14: 21102455-21102455 on Build GRCh38
rs565943809 Chr.14: 21102447-21102447 on Build GRCh38
rs925314857 Chr.14: 21102395-21102395 on Build GRCh38
rs535456111 Chr.14: 21102326-21102326 on Build GRCh38
rs553887169 Chr.14: 21102468-21102468 on Build GRCh38
rs781660269 Chr.14: 21102408-21102408 on Build GRCh38
rs534149594 Chr.14: 21102467-21102467 on Build GRCh38
rs537729067 Chr.14: 21102411-21102411 on Build GRCh38
rs760683015 Chr.14: 21102462-21102462 on Build GRCh38
rs747587357 Chr.14: 21102390-21102390 on Build GRCh38
rs963995244 Chr.14: 21102346-21102346 on Build GRCh38
rs546027393 Chr.14: 21102534-21102534 on Build GRCh38
rs759131097 Chr.14: 21102421-21102421 on Build GRCh38
rs775454146 Chr.14: 21102429-21102429 on Build GRCh38
rs1045321026 Chr.14: 21102493-21102493 on Build GRCh38
rs926904319 Chr.14: 21102501-21102501 on Build GRCh38
rs554406900 Chr.14: 21102423-21102423 on Build GRCh38
rs957704312 Chr.14: 21102426-21102426 on Build GRCh38
rs577258726 Chr.14: 21102483-21102483 on Build GRCh38
rs966410483 Chr.14: 21102464-21102464 on Build GRCh38
rs1018175783 Chr.14: 21102337-21102337 on Build GRCh38

View Full Product Details