Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99662707-99662978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A5
Entrez Gene ID
1577
Gene Name
cytochrome P450 family 3 subfamily A member 5
Gene Aliases
CP35, CYPIIIA5, P450PCN3, PCN3
UniGene
Hs.571258
Gene Chromosome Location
Chr.7: 99648189-99680026 on Build GRCh38
Transcripts
NM_000777.4 NM_001291829.1 NM_001291830.1 NR_033807.2 XM_006715859.2 XM_011515843.1 XM_017011781.1 XM_017011782.1 XM_011515909.2 XM_017011827.1

Gene Symbol
ZSCAN25
Entrez Gene ID
221785
Gene Name
zinc finger and SCAN domain containing 25
Gene Aliases
ZNF498
UniGene
Hs.446297
Gene Chromosome Location
Chr.7: 99616600-99738689 on Build GRCh38
Transcripts
NM_000777.4 NM_001291829.1 NM_001291830.1 NR_033807.2 XM_006715859.2 XM_011515843.1 XM_017011781.1 XM_017011782.1 XM_011515909.2 XM_017011827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6481914 c.792G>A p.L264L Chr.7: 99662859-99662859 on Build GRCh38
7202758 c.803G>A p.R268Q Chr.7: 99662878-99662878 on Build GRCh38
7422883 c.833C>A p.S278Y Chr.7: 99662848-99662848 on Build GRCh38
4993037 c.809A>G p.N270S Chr.7: 99662842-99662842 on Build GRCh38
7108668 c.787C>A p.Q263K Chr.7: 99662864-99662864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148176345 Chr.7: 99662879-99662879 on Build GRCh38
rs188843385 Chr.7: 99662888-99662888 on Build GRCh38
rs28365096 Chr.7: 99662876-99662876 on Build GRCh38
rs753396001 Chr.7: 99662857-99662857 on Build GRCh38
rs770099217 Chr.7: 99662804-99662804 on Build GRCh38
rs372459335 Chr.7: 99662924-99662924 on Build GRCh38
rs963831892 Chr.7: 99662941-99662941 on Build GRCh38
rs751478483 Chr.7: 99662776-99662776 on Build GRCh38
rs912206185 Chr.7: 99662738-99662738 on Build GRCh38
rs777973485 Chr.7: 99662920-99662920 on Build GRCh38
rs538867950 Chr.7: 99662793-99662793 on Build GRCh38
rs749311811 Chr.7: 99662923-99662923 on Build GRCh38
rs1005318700 Chr.7: 99662910-99662910 on Build GRCh38
rs376884726 Chr.7: 99662783-99662783 on Build GRCh38
rs202135035 Chr.7: 99662863-99662863 on Build GRCh38
rs28383478 Chr.7: 99662784-99662784 on Build GRCh38
rs58708491 Chr.7: 99662909-99662909 on Build GRCh38
rs763084061 Chr.7: 99662823-99662823 on Build GRCh38
rs145774441 Chr.7: 99662854-99662854 on Build GRCh38
rs974861077 Chr.7: 99662942-99662942 on Build GRCh38
rs111371159 Chr.7: 99662839-99662839 on Build GRCh38
rs766943983 Chr.7: 99662791-99662791 on Build GRCh38
rs771363355 Chr.7: 99662826-99662826 on Build GRCh38
rs199896750 Chr.7: 99662851-99662851 on Build GRCh38
rs4646453 Chr.7: 99662739-99662739 on Build GRCh38
rs900353973 Chr.7: 99662779-99662779 on Build GRCh38
rs759899966 Chr.7: 99662838-99662838 on Build GRCh38
rs779989195 Chr.7: 99662878-99662878 on Build GRCh38
rs774855888 Chr.7: 99662828-99662828 on Build GRCh38
rs755819922 Chr.7: 99662902-99662902 on Build GRCh38
rs28383477 Chr.7: 99662850-99662850 on Build GRCh38
rs773263948 Chr.7: 99662812-99662812 on Build GRCh38
rs1044466773 Chr.7: 99662949-99662949 on Build GRCh38
rs756839053 Chr.7: 99662860-99662860 on Build GRCh38
rs771024087 Chr.7: 99662932-99662932 on Build GRCh38
rs546020107 Chr.7: 99662954-99662980 on Build GRCh38
rs769901021 Chr.7: 99662912-99662912 on Build GRCh38

View Full Product Details