Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 219971978-219972241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPRS
Entrez Gene ID
2058
Gene Name
glutamyl-prolyl-tRNA synthetase
Gene Aliases
EARS, GLUPRORS, PARS, PIG32, QARS, QPRS
UniGene
Hs.497788
Gene Chromosome Location
Chr.1: 219968600-220046658 on Build GRCh38
Transcripts
NM_004446.2 XM_017000614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6460069 c.4300C>T p.Q1434* Chr.1: 219972092-219972092 on Build GRCh38
6061058 c.4250C>T p.S1417F Chr.1: 219972142-219972142 on Build GRCh38
6429016 c.4279G>T p.A1427S Chr.1: 219972113-219972113 on Build GRCh38
4028358 c.4277T>C p.V1426A Chr.1: 219972115-219972115 on Build GRCh38
5546327 c.4246G>A p.A1416T Chr.1: 219972146-219972146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757886414 Chr.1: 219972168-219972168 on Build GRCh38
rs752157288 Chr.1: 219972162-219972162 on Build GRCh38
rs533671119 Chr.1: 219972202-219972202 on Build GRCh38
rs776132512 Chr.1: 219972047-219972047 on Build GRCh38
rs754808226 Chr.1: 219972120-219972120 on Build GRCh38
rs774975979 Chr.1: 219972187-219972187 on Build GRCh38
rs763357920 Chr.1: 219972050-219972050 on Build GRCh38
rs528835769 Chr.1: 219972085-219972085 on Build GRCh38
rs765490624 Chr.1: 219972103-219972107 on Build GRCh38
rs1013887606 Chr.1: 219972186-219972186 on Build GRCh38
rs780226725 Chr.1: 219972179-219972179 on Build GRCh38
rs778518963 Chr.1: 219972145-219972145 on Build GRCh38
rs999916677 Chr.1: 219972060-219972061 on Build GRCh38
rs375120694 Chr.1: 219972182-219972182 on Build GRCh38
rs768994588 Chr.1: 219972183-219972183 on Build GRCh38
rs1041594662 Chr.1: 219972028-219972028 on Build GRCh38
rs746808658 Chr.1: 219972076-219972076 on Build GRCh38
rs746281647 Chr.1: 219972033-219972033 on Build GRCh38
rs771676146 Chr.1: 219972020-219972020 on Build GRCh38
rs746660197 Chr.1: 219972174-219972174 on Build GRCh38
rs777441821 Chr.1: 219972170-219972170 on Build GRCh38
rs559389537 Chr.1: 219972092-219972092 on Build GRCh38
rs548851329 Chr.1: 219972091-219972091 on Build GRCh38
rs1007236484 Chr.1: 219972147-219972147 on Build GRCh38
rs770252533 Chr.1: 219972175-219972175 on Build GRCh38
rs766071770 Chr.1: 219972101-219972101 on Build GRCh38
rs764236465 Chr.1: 219972075-219972075 on Build GRCh38
rs757021003 Chr.1: 219972064-219972064 on Build GRCh38
rs753825531 Chr.1: 219972105-219972105 on Build GRCh38
rs148671123 Chr.1: 219972083-219972083 on Build GRCh38
rs755058707 Chr.1: 219972041-219972041 on Build GRCh38
rs371986872 Chr.1: 219972146-219972146 on Build GRCh38
rs2795315 Chr.1: 219972029-219972029 on Build GRCh38
rs527889239 Chr.1: 219972202-219972202 on Build GRCh38
rs2795315 Chr.1: 219972029-219972029 on Build GRCh38
rs543027877 Chr.1: 219972046-219972046 on Build GRCh38

View Full Product Details