Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3905255-3905528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
UniGene
Hs.418055
Gene Chromosome Location
Chr.19: 3880620-3928082 on Build GRCh38
Transcripts
NM_033064.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3797041 c.163G>T p.G55* Chr.19: 3905460-3905460 on Build GRCh38
711594 c.159C>T p.F53F Chr.19: 3905456-3905456 on Build GRCh38
4533605 c.199G>A p.E67K Chr.19: 3905496-3905496 on Build GRCh38
5904474 c.137-7C>T p.? Chr.19: 3905427-3905427 on Build GRCh38
3222628 c.162C>T p.N54N Chr.19: 3905459-3905459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554981913 Chr.19: 3905405-3905405 on Build GRCh38
rs764109827 Chr.19: 3905441-3905441 on Build GRCh38
rs776023908 Chr.19: 3905506-3905506 on Build GRCh38
rs373392142 Chr.19: 3905459-3905459 on Build GRCh38
rs774698649 Chr.19: 3905422-3905422 on Build GRCh38
rs762132813 Chr.19: 3905447-3905447 on Build GRCh38
rs943105824 Chr.19: 3905376-3905376 on Build GRCh38
rs754056923 Chr.19: 3905446-3905446 on Build GRCh38
rs772361307 Chr.19: 3905498-3905498 on Build GRCh38
rs538402960 Chr.19: 3905398-3905398 on Build GRCh38
rs768054237 Chr.19: 3905426-3905426 on Build GRCh38
rs934320237 Chr.19: 3905308-3905308 on Build GRCh38
rs983461799 Chr.19: 3905411-3905411 on Build GRCh38
rs1040164496 Chr.19: 3905287-3905287 on Build GRCh38
rs375319290 Chr.19: 3905283-3905283 on Build GRCh38
rs951648133 Chr.19: 3905401-3905401 on Build GRCh38
rs1051393169 Chr.19: 3905348-3905348 on Build GRCh38
rs370499062 Chr.19: 3905482-3905482 on Build GRCh38
rs751387636 Chr.19: 3905398-3905398 on Build GRCh38
rs917266568 Chr.19: 3905378-3905378 on Build GRCh38
rs775910010 Chr.19: 3905431-3905431 on Build GRCh38
rs778595221 Chr.19: 3905483-3905483 on Build GRCh38
rs199582357 Chr.19: 3905464-3905464 on Build GRCh38
rs376454003 Chr.19: 3905469-3905469 on Build GRCh38
rs111345257 Chr.19: 3905292-3905292 on Build GRCh38
rs771224365 Chr.19: 3905399-3905399 on Build GRCh38
rs73919387 Chr.19: 3905423-3905423 on Build GRCh38
rs567549652 Chr.19: 3905389-3905390 on Build GRCh38
rs1000284091 Chr.19: 3905307-3905307 on Build GRCh38
rs958786035 Chr.19: 3905341-3905341 on Build GRCh38
rs933763316 Chr.19: 3905458-3905458 on Build GRCh38
rs781157054 Chr.19: 3905460-3905460 on Build GRCh38
rs757164887 Chr.19: 3905407-3905407 on Build GRCh38
rs1034467558 Chr.19: 3905325-3905325 on Build GRCh38
rs369506043 Chr.19: 3905440-3905440 on Build GRCh38
rs761591994 Chr.19: 3905356-3905356 on Build GRCh38
rs535876762 Chr.19: 3905449-3905449 on Build GRCh38
rs748869597 Chr.19: 3905472-3905472 on Build GRCh38
rs549284827 Chr.19: 3905293-3905293 on Build GRCh38
rs746208351 Chr.19: 3905495-3905495 on Build GRCh38
rs776843396 Chr.19: 3905323-3905323 on Build GRCh38
rs745666579 Chr.19: 3905305-3905305 on Build GRCh38
rs199662587 Chr.19: 3905470-3905470 on Build GRCh38

View Full Product Details