Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119372144-119372417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBI
Entrez Gene ID
1622
Gene Name
diazepam binding inhibitor, acyl-CoA binding protein
Gene Aliases
ACBD1, ACBP, CCK-RP, EP
UniGene
Hs.78888
Gene Chromosome Location
Chr.2: 119366924-119372551 on Build GRCh38
Transcripts
NM_001079862.2 NM_001079863.1 NM_001178017.1 NM_001178041.2 NM_001178042.2 NM_001178043.2 NM_001282633.1 NM_001282634.1 NM_001282635.1 NM_001282636.1 NM_020548.7 NR_104221.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991109 c.444A>G p.I148M Chr.2: 119372315-119372315 on Build GRCh38
475848 c.242-2A>T p.? Chr.2: 119372243-119372243 on Build GRCh38
3836684 c.429G>T p.K143N Chr.2: 119372300-119372300 on Build GRCh38
1005922 c.299A>C p.K100T Chr.2: 119372302-119372302 on Build GRCh38
1399054 c.297G>T p.K99N Chr.2: 119372300-119372300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768833942 Chr.2: 119372194-119372194 on Build GRCh38
rs138712234 Chr.2: 119372311-119372311 on Build GRCh38
rs200142643 Chr.2: 119372357-119372357 on Build GRCh38
rs201004978 Chr.2: 119372309-119372309 on Build GRCh38
rs368735097 Chr.2: 119372338-119372338 on Build GRCh38
rs8192506 Chr.2: 119372265-119372265 on Build GRCh38
rs771529766 Chr.2: 119372244-119372244 on Build GRCh38
rs777846902 Chr.2: 119372353-119372353 on Build GRCh38
rs375121791 Chr.2: 119372301-119372301 on Build GRCh38
rs1803482 Chr.2: 119372380-119372380 on Build GRCh38
rs763621561 Chr.2: 119372288-119372288 on Build GRCh38
rs1051232530 Chr.2: 119372375-119372375 on Build GRCh38
rs964270228 Chr.2: 119372258-119372258 on Build GRCh38
rs879437677 Chr.2: 119372214-119372214 on Build GRCh38
rs8192507 Chr.2: 119372310-119372310 on Build GRCh38
rs917081074 Chr.2: 119372260-119372260 on Build GRCh38
rs774821188 Chr.2: 119372246-119372246 on Build GRCh38
rs777381954 Chr.2: 119372370-119372370 on Build GRCh38
rs368008808 Chr.2: 119372204-119372204 on Build GRCh38
rs112491476 Chr.2: 119372298-119372298 on Build GRCh38
rs567360937 Chr.2: 119372187-119372187 on Build GRCh38
rs776066590 Chr.2: 119372289-119372289 on Build GRCh38
rs1803481 Chr.2: 119372278-119372278 on Build GRCh38
rs766387683 Chr.2: 119372313-119372313 on Build GRCh38
rs200533076 Chr.2: 119372257-119372257 on Build GRCh38
rs771173404 Chr.2: 119372358-119372358 on Build GRCh38
rs751511709 Chr.2: 119372319-119372319 on Build GRCh38
rs749713771 Chr.2: 119372355-119372355 on Build GRCh38
rs878978443 Chr.2: 119372240-119372240 on Build GRCh38
rs879145893 Chr.2: 119372242-119372242 on Build GRCh38
rs758114328 Chr.2: 119372312-119372312 on Build GRCh38
rs754915009 Chr.2: 119372326-119372326 on Build GRCh38
rs192533113 Chr.2: 119372390-119372390 on Build GRCh38
rs867162287 Chr.2: 119372351-119372351 on Build GRCh38
rs756426832 Chr.2: 119372344-119372344 on Build GRCh38
rs372038078 Chr.2: 119372233-119372233 on Build GRCh38
rs781381032 Chr.2: 119372202-119372202 on Build GRCh38
rs769852138 Chr.2: 119372223-119372223 on Build GRCh38
rs188178195 Chr.2: 119372331-119372331 on Build GRCh38
rs748173514 Chr.2: 119372342-119372342 on Build GRCh38

View Full Product Details