Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54362688-54362955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
UniGene
Hs.208701
Gene Chromosome Location
Chr.18: 54357914-54382035 on Build GRCh38
Transcripts
NM_001288980.1 NM_001288981.1 NM_001288982.1 NM_173529.5 NR_110234.1 XM_005258201.1 XM_005258203.1 XM_017025567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821580 c.719G>T p.W240L Chr.18: 54362900-54362900 on Build GRCh38
6564604 c.615G>A p.L205L Chr.18: 54362796-54362796 on Build GRCh38
6430863 c.689C>A p.S230* Chr.18: 54362870-54362870 on Build GRCh38
988988 c.626G>T p.R209I Chr.18: 54362807-54362807 on Build GRCh38
5682437 c.736A>C p.T246P Chr.18: 54362917-54362917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143596631 Chr.18: 54362801-54362801 on Build GRCh38
rs868213847 Chr.18: 54362920-54362921 on Build GRCh38
rs780006933 Chr.18: 54362811-54362811 on Build GRCh38
rs748312524 Chr.18: 54362812-54362812 on Build GRCh38
rs74862391 Chr.18: 54362887-54362887 on Build GRCh38
rs775839798 Chr.18: 54362850-54362850 on Build GRCh38
rs760532831 Chr.18: 54362898-54362898 on Build GRCh38
rs758757569 Chr.18: 54362814-54362814 on Build GRCh38
rs750355302 Chr.18: 54362795-54362795 on Build GRCh38
rs140618781 Chr.18: 54362892-54362892 on Build GRCh38
rs771312594 Chr.18: 54362839-54362839 on Build GRCh38
rs567692537 Chr.18: 54362824-54362824 on Build GRCh38
rs757117440 Chr.18: 54362788-54362788 on Build GRCh38
rs904551781 Chr.18: 54362810-54362810 on Build GRCh38
rs746388601 Chr.18: 54362845-54362845 on Build GRCh38
rs553514585 Chr.18: 54362822-54362822 on Build GRCh38
rs766954356 Chr.18: 54362873-54362873 on Build GRCh38
rs766270802 Chr.18: 54362905-54362905 on Build GRCh38
rs775078661 Chr.18: 54362746-54362746 on Build GRCh38
rs763530149 Chr.18: 54362766-54362766 on Build GRCh38
rs369589214 Chr.18: 54362860-54362860 on Build GRCh38
rs759985779 Chr.18: 54362726-54362726 on Build GRCh38
rs781695334 Chr.18: 54362840-54362840 on Build GRCh38
rs1000420860 Chr.18: 54362844-54362844 on Build GRCh38
rs762592758 Chr.18: 54362858-54362858 on Build GRCh38
rs767526321 Chr.18: 54362791-54362791 on Build GRCh38
rs1015033517 Chr.18: 54362908-54362908 on Build GRCh38
rs895833568 Chr.18: 54362869-54362869 on Build GRCh38
rs778306204 Chr.18: 54362922-54362925 on Build GRCh38
rs765556175 Chr.18: 54362745-54362745 on Build GRCh38
rs144173544 Chr.18: 54362918-54362918 on Build GRCh38
rs770141925 Chr.18: 54362848-54362848 on Build GRCh38
rs773816620 Chr.18: 54362867-54362867 on Build GRCh38
rs762611126 Chr.18: 54362751-54362751 on Build GRCh38
rs932790139 Chr.18: 54362728-54362728 on Build GRCh38
rs778182138 Chr.18: 54362922-54362922 on Build GRCh38
rs748333505 Chr.18: 54362783-54362783 on Build GRCh38
rs1050173289 Chr.18: 54362767-54362767 on Build GRCh38
rs201042542 Chr.18: 54362917-54362917 on Build GRCh38

View Full Product Details