Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41938974-41939245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPM
Entrez Gene ID
79019
Gene Name
centromere protein M
Gene Aliases
C22orf18, CENP-M, PANE1
UniGene
Hs.208912
Gene Chromosome Location
Chr.22: 41927747-41947164 on Build GRCh38
Transcripts
NM_001002876.2 NM_001110215.2 NM_001304370.1 NM_001304372.1 NM_001304373.1 NM_024053.4 XM_011530368.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7312791 c.108C>A p.P36P Chr.22: 41939125-41939125 on Build GRCh38
4634807 c.432G>A p.Q144Q Chr.22: 41939167-41939167 on Build GRCh38
6626651 c.129G>A p.L43L Chr.22: 41939104-41939104 on Build GRCh38
6004943 c.63G>C p.A21A Chr.22: 41939170-41939170 on Build GRCh38
5393762 c.500C>T p.S167F Chr.22: 41939099-41939099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762543264 Chr.22: 41939208-41939208 on Build GRCh38
rs755984435 Chr.22: 41939226-41939226 on Build GRCh38
rs756847529 Chr.22: 41939132-41939132 on Build GRCh38
rs762296545 Chr.22: 41939172-41939181 on Build GRCh38
rs374540211 Chr.22: 41939157-41939157 on Build GRCh38
rs755387483 Chr.22: 41939018-41939018 on Build GRCh38
rs764776097 Chr.22: 41939205-41939205 on Build GRCh38
rs774949456 Chr.22: 41939120-41939121 on Build GRCh38
rs771006490 Chr.22: 41939196-41939196 on Build GRCh38
rs376664319 Chr.22: 41939171-41939171 on Build GRCh38
rs570248268 Chr.22: 41939064-41939064 on Build GRCh38
rs756942871 Chr.22: 41939038-41939038 on Build GRCh38
rs757973961 Chr.22: 41939146-41939146 on Build GRCh38
rs184253320 Chr.22: 41939197-41939197 on Build GRCh38
rs945161784 Chr.22: 41939093-41939093 on Build GRCh38
rs748314408 Chr.22: 41939074-41939074 on Build GRCh38
rs765008124 Chr.22: 41939141-41939141 on Build GRCh38
rs773521621 Chr.22: 41939175-41939175 on Build GRCh38
rs776598765 Chr.22: 41939069-41939069 on Build GRCh38
rs546315747 Chr.22: 41939004-41939004 on Build GRCh38
rs143638439 Chr.22: 41939152-41939152 on Build GRCh38
rs138055602 Chr.22: 41939143-41939143 on Build GRCh38
rs763157368 Chr.22: 41939106-41939106 on Build GRCh38
rs774792724 Chr.22: 41939124-41939124 on Build GRCh38
rs766762973 Chr.22: 41939113-41939113 on Build GRCh38
rs376708928 Chr.22: 41939201-41939201 on Build GRCh38
rs749496721 Chr.22: 41939176-41939176 on Build GRCh38
rs569652175 Chr.22: 41939125-41939125 on Build GRCh38
rs1015638569 Chr.22: 41938997-41938997 on Build GRCh38
rs368867350 Chr.22: 41939033-41939033 on Build GRCh38
rs763306762 Chr.22: 41939011-41939011 on Build GRCh38
rs781341914 Chr.22: 41939221-41939221 on Build GRCh38
rs373230417 Chr.22: 41939044-41939044 on Build GRCh38
rs773148657 Chr.22: 41939095-41939095 on Build GRCh38
rs764711908 Chr.22: 41939111-41939112 on Build GRCh38
rs760069349 Chr.22: 41939130-41939130 on Build GRCh38
rs775132155 Chr.22: 41939022-41939038 on Build GRCh38
rs751127633 Chr.22: 41939213-41939213 on Build GRCh38
rs767807033 Chr.22: 41939131-41939131 on Build GRCh38
rs754948842 Chr.22: 41939220-41939220 on Build GRCh38
rs534722619 Chr.22: 41939209-41939209 on Build GRCh38
rs901118819 Chr.22: 41939000-41939000 on Build GRCh38
rs770064281 Chr.22: 41939085-41939085 on Build GRCh38
rs894941464 Chr.22: 41939150-41939150 on Build GRCh38
rs781072264 Chr.22: 41939165-41939165 on Build GRCh38
rs750063789 Chr.22: 41939166-41939166 on Build GRCh38
rs914989724 Chr.22: 41939087-41939087 on Build GRCh38
rs762484483 Chr.22: 41939066-41939067 on Build GRCh38
rs376167757 Chr.22: 41939017-41939017 on Build GRCh38
rs533074775 Chr.22: 41938996-41938996 on Build GRCh38
rs750042043 Chr.22: 41939207-41939207 on Build GRCh38
rs549021345 Chr.22: 41939170-41939170 on Build GRCh38
rs994685502 Chr.22: 41939154-41939154 on Build GRCh38
rs371185369 Chr.22: 41939156-41939156 on Build GRCh38
rs746777274 Chr.22: 41939061-41939061 on Build GRCh38
rs1049408107 Chr.22: 41939155-41939155 on Build GRCh38
rs977047627 Chr.22: 41939023-41939023 on Build GRCh38
rs750517475 Chr.22: 41939010-41939010 on Build GRCh38
rs767893562 Chr.22: 41939027-41939027 on Build GRCh38
rs761142735 Chr.22: 41939204-41939204 on Build GRCh38
rs541706503 Chr.22: 41939200-41939200 on Build GRCh38
rs780183010 Chr.22: 41939060-41939060 on Build GRCh38
rs757994312 Chr.22: 41939192-41939192 on Build GRCh38
rs377466334 Chr.22: 41939045-41939045 on Build GRCh38
rs765189565 Chr.22: 41939006-41939006 on Build GRCh38

View Full Product Details