Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150730817-150731090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN4
Entrez Gene ID
51164
Gene Name
dynactin subunit 4
Gene Aliases
DYN4, P62
UniGene
Hs.675564
Gene Chromosome Location
Chr.5: 150708747-150759095 on Build GRCh38
Transcripts
NM_001135643.1 NM_001135644.1 NM_016221.3 XM_005268457.1 XM_011537644.1 XM_011537645.1 XM_011537646.1 XM_011537647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1157741 c.707C>A p.P236Q Chr.5: 150731061-150731061 on Build GRCh38
4676629 c.699T>C p.Y233Y Chr.5: 150731069-150731069 on Build GRCh38
3784662 c.721G>A p.E241K Chr.5: 150731047-150731047 on Build GRCh38
6356573 c.682C>A p.L228I Chr.5: 150731086-150731086 on Build GRCh38
7199001 c.687T>G p.P229P Chr.5: 150731081-150731081 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965528368 Chr.5: 150730883-150730883 on Build GRCh38
rs574100804 Chr.5: 150730864-150730864 on Build GRCh38
rs77008114 Chr.5: 150730853-150730853 on Build GRCh38
rs376493797 Chr.5: 150731003-150731003 on Build GRCh38
rs377678851 Chr.5: 150730956-150730956 on Build GRCh38
rs753376523 Chr.5: 150730872-150730872 on Build GRCh38
rs1006338233 Chr.5: 150730897-150730897 on Build GRCh38
rs761589223 Chr.5: 150731005-150731005 on Build GRCh38
rs1017758189 Chr.5: 150730916-150730916 on Build GRCh38
rs964913778 Chr.5: 150730930-150730930 on Build GRCh38
rs925381364 Chr.5: 150730978-150730978 on Build GRCh38
rs763366168 Chr.5: 150731060-150731060 on Build GRCh38
rs252154 Chr.5: 150730848-150730848 on Build GRCh38
rs563292405 Chr.5: 150730873-150730873 on Build GRCh38
rs989002749 Chr.5: 150730904-150730904 on Build GRCh38
rs779933769 Chr.5: 150730936-150730938 on Build GRCh38
rs969346884 Chr.5: 150730950-150730950 on Build GRCh38
rs115544229 Chr.5: 150730936-150730936 on Build GRCh38
rs779552884 Chr.5: 150730952-150730952 on Build GRCh38
rs766970015 Chr.5: 150731061-150731061 on Build GRCh38
rs772673495 Chr.5: 150730986-150730986 on Build GRCh38
rs748568126 Chr.5: 150730981-150730981 on Build GRCh38
rs778382204 Chr.5: 150731058-150731058 on Build GRCh38
rs978535040 Chr.5: 150730970-150730970 on Build GRCh38
rs769377612 Chr.5: 150731011-150731011 on Build GRCh38
rs772861285 Chr.5: 150731016-150731016 on Build GRCh38
rs887563521 Chr.5: 150730888-150730891 on Build GRCh38

View Full Product Details