Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 884015-884254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHID1
Entrez Gene ID
66005
Gene Name
chitinase domain containing 1
Gene Aliases
GL008, SI-CLP, SICLP
UniGene
Hs.144468
Gene Chromosome Location
Chr.11: 867859-915058 on Build GRCh38
Transcripts
NM_001142674.1 NM_001142675.1 NM_001142676.1 NM_001142677.1 NM_023947.3 XM_005253072.2 XM_005253073.4 XM_006718282.2 XM_011520299.1 XM_011520300.1 XM_011520301.2 XM_011520302.2 XM_017018166.1 XM_017018167.1 XM_017018168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453505 c.873C>T p.Y291Y Chr.11: 884088-884088 on Build GRCh38
4381860 c.810G>T p.M270I Chr.11: 884151-884151 on Build GRCh38
1731345 c.706G>T p.D236Y Chr.11: 884165-884165 on Build GRCh38
4868193 c.819C>T p.H273H Chr.11: 884142-884142 on Build GRCh38
6437212 c.847C>T p.L283L Chr.11: 884114-884114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763000425 Chr.11: 884145-884145 on Build GRCh38
rs770223032 Chr.11: 884123-884123 on Build GRCh38
rs750308081 Chr.11: 884076-884076 on Build GRCh38
rs778245299 Chr.11: 884037-884037 on Build GRCh38
rs771249083 Chr.11: 884042-884042 on Build GRCh38
rs753432580 Chr.11: 884088-884088 on Build GRCh38
rs536210286 Chr.11: 884116-884116 on Build GRCh38
rs760623311 Chr.11: 884172-884172 on Build GRCh38
rs751831737 Chr.11: 884061-884061 on Build GRCh38
rs756019950 Chr.11: 884078-884078 on Build GRCh38
rs149001064 Chr.11: 884151-884151 on Build GRCh38
rs569770877 Chr.11: 884175-884175 on Build GRCh38
rs781411027 Chr.11: 884120-884120 on Build GRCh38
rs373822870 Chr.11: 884180-884180 on Build GRCh38
rs763884284 Chr.11: 884048-884048 on Build GRCh38
rs112897109 Chr.11: 884065-884065 on Build GRCh38
rs113539115 Chr.11: 884066-884066 on Build GRCh38
rs773964018 Chr.11: 884189-884189 on Build GRCh38
rs781567580 Chr.11: 884216-884216 on Build GRCh38
rs368689362 Chr.11: 884207-884207 on Build GRCh38
rs899315017 Chr.11: 884182-884182 on Build GRCh38
rs760023779 Chr.11: 884045-884045 on Build GRCh38
rs763110955 Chr.11: 884054-884054 on Build GRCh38
rs574373739 Chr.11: 884233-884233 on Build GRCh38
rs756521843 Chr.11: 884221-884221 on Build GRCh38
rs750208434 Chr.11: 884169-884169 on Build GRCh38
rs775506286 Chr.11: 884126-884126 on Build GRCh38
rs146861158 Chr.11: 884146-884146 on Build GRCh38
rs779663358 Chr.11: 884087-884087 on Build GRCh38
rs775557617 Chr.11: 884052-884052 on Build GRCh38
rs561006487 Chr.11: 884226-884226 on Build GRCh38
rs145103748 Chr.11: 884139-884139 on Build GRCh38
rs61869682 Chr.11: 884193-884193 on Build GRCh38
rs905315937 Chr.11: 884163-884163 on Build GRCh38
rs886775112 Chr.11: 884204-884204 on Build GRCh38
rs767285671 Chr.11: 884074-884074 on Build GRCh38
rs201116368 Chr.11: 884160-884160 on Build GRCh38
rs754520811 Chr.11: 884185-884185 on Build GRCh38
rs567536055 Chr.11: 884057-884057 on Build GRCh38
rs377704138 Chr.11: 884219-884219 on Build GRCh38
rs757996138 Chr.11: 884205-884205 on Build GRCh38
rs143837907 Chr.11: 884165-884165 on Build GRCh38
rs754573713 Chr.11: 884103-884103 on Build GRCh38
rs376597827 Chr.11: 884117-884117 on Build GRCh38
rs777182034 Chr.11: 884044-884044 on Build GRCh38
rs767285054 Chr.11: 884166-884166 on Build GRCh38
rs747345472 Chr.11: 884040-884040 on Build GRCh38
rs769841632 Chr.11: 884049-884049 on Build GRCh38
rs141328460 Chr.11: 884118-884118 on Build GRCh38
rs753873744 Chr.11: 884183-884183 on Build GRCh38
rs762680331 Chr.11: 884047-884047 on Build GRCh38
rs375089194 Chr.11: 884073-884073 on Build GRCh38
rs892630683 Chr.11: 884210-884210 on Build GRCh38
rs746324429 Chr.11: 884122-884122 on Build GRCh38

View Full Product Details