Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54588646-54588901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK5
Entrez Gene ID
55816
Gene Name
docking protein 5
Gene Aliases
C20orf180, IRS-6, IRS6
UniGene
Hs.473133
Gene Chromosome Location
Chr.20: 54475472-54651171 on Build GRCh38
Transcripts
NM_018431.4 NM_177959.2 XM_011528904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444070 c.290-1G>T p.? Chr.20: 54588686-54588686 on Build GRCh38
6866888 c.400G>A p.E134K Chr.20: 54588797-54588797 on Build GRCh38
4968618 c.343_349delCGGATCA p.R115fs*27 Chr.20: 54588740-54588746 on Build GRCh38
6481323 c.336A>G p.V112V Chr.20: 54588733-54588733 on Build GRCh38
3963680 c.373G>C p.D125H Chr.20: 54588770-54588770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753959704 Chr.20: 54588804-54588804 on Build GRCh38
rs767805238 Chr.20: 54588699-54588699 on Build GRCh38
rs879937546 Chr.20: 54588810-54588810 on Build GRCh38
rs766427445 Chr.20: 54588843-54588843 on Build GRCh38
rs16999812 Chr.20: 54588742-54588742 on Build GRCh38
rs764290120 Chr.20: 54588708-54588708 on Build GRCh38
rs756349198 Chr.20: 54588701-54588701 on Build GRCh38
rs552375662 Chr.20: 54588707-54588707 on Build GRCh38
rs757463502 Chr.20: 54588713-54588713 on Build GRCh38
rs759981209 Chr.20: 54588764-54588764 on Build GRCh38
rs1002185419 Chr.20: 54588866-54588866 on Build GRCh38
rs916108882 Chr.20: 54588788-54588788 on Build GRCh38
rs1052669335 Chr.20: 54588760-54588760 on Build GRCh38
rs1028805290 Chr.20: 54588677-54588677 on Build GRCh38
rs779157197 Chr.20: 54588715-54588715 on Build GRCh38
rs764355081 Chr.20: 54588793-54588793 on Build GRCh38
rs112545680 Chr.20: 54588726-54588726 on Build GRCh38
rs757586570 Chr.20: 54588809-54588809 on Build GRCh38
rs754969106 Chr.20: 54588729-54588729 on Build GRCh38
rs768376773 Chr.20: 54588728-54588728 on Build GRCh38
rs375379923 Chr.20: 54588827-54588827 on Build GRCh38
rs768075273 Chr.20: 54588770-54588770 on Build GRCh38
rs935022234 Chr.20: 54588863-54588863 on Build GRCh38
rs753923771 Chr.20: 54588710-54588710 on Build GRCh38
rs760054932 Chr.20: 54588693-54588693 on Build GRCh38
rs760986080 Chr.20: 54588785-54588785 on Build GRCh38
rs150360240 Chr.20: 54588748-54588748 on Build GRCh38
rs200020780 Chr.20: 54588750-54588750 on Build GRCh38
rs752108025 Chr.20: 54588681-54588681 on Build GRCh38
rs6014067 Chr.20: 54588857-54588857 on Build GRCh38
rs773975138 Chr.20: 54588740-54588740 on Build GRCh38
rs148783714 Chr.20: 54588741-54588741 on Build GRCh38
rs1011030736 Chr.20: 54588794-54588794 on Build GRCh38
rs746902095 Chr.20: 54588716-54588716 on Build GRCh38
rs775922103 Chr.20: 54588782-54588782 on Build GRCh38
rs201026396 Chr.20: 54588799-54588799 on Build GRCh38
rs748807469 Chr.20: 54588747-54588747 on Build GRCh38
rs199704788 Chr.20: 54588730-54588730 on Build GRCh38
rs763208647 Chr.20: 54588709-54588709 on Build GRCh38
rs979024815 Chr.20: 54588835-54588835 on Build GRCh38
rs183251839 Chr.20: 54588834-54588834 on Build GRCh38

View Full Product Details