Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97589756-97590029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf62
Entrez Gene ID
414157
Gene Name
chromosome 10 open reading frame 62
Gene Aliases
bA548K23.1
UniGene
Hs.662302
Gene Chromosome Location
Chr.10: 97589693-97590934 on Build GRCh38
Transcripts
NM_001009997.2 NM_001134670.1 NM_138413.3

Gene Symbol
HOGA1
Entrez Gene ID
112817
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
C10orf65, DHDPS2, DHDPSL, HP3, NPL2
UniGene
Hs.180346
Gene Chromosome Location
Chr.10: 97584345-97612802 on Build GRCh38
Transcripts
NM_001009997.2 NM_001134670.1 NM_138413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613883 c.122G>A p.R41H Chr.10: 97590019-97590019 on Build GRCh38
3368178 c.21G>C p.K7N Chr.10: 97589918-97589918 on Build GRCh38
4666524 c.62A>G p.D21G Chr.10: 97589959-97589959 on Build GRCh38
3985841 c.40T>A p.S14T Chr.10: 97589937-97589937 on Build GRCh38
1492425 c.51A>T p.P17P Chr.10: 97589948-97589948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540202969 Chr.10: 97589839-97589839 on Build GRCh38
rs149631769 Chr.10: 97589926-97589926 on Build GRCh38
rs568126358 Chr.10: 97589846-97589846 on Build GRCh38
rs931517408 Chr.10: 97589798-97589798 on Build GRCh38
rs770023306 Chr.10: 97589900-97589900 on Build GRCh38
rs908811315 Chr.10: 97589816-97589816 on Build GRCh38
rs11540693 Chr.10: 97589781-97589781 on Build GRCh38
rs749423744 Chr.10: 97589918-97589920 on Build GRCh38
rs753388582 Chr.10: 97589977-97589977 on Build GRCh38
rs777463197 Chr.10: 97589965-97589965 on Build GRCh38
rs779224982 Chr.10: 97589841-97589841 on Build GRCh38
rs748267007 Chr.10: 97589953-97589953 on Build GRCh38
rs767480560 Chr.10: 97589952-97589952 on Build GRCh38
rs778331591 Chr.10: 97589982-97589982 on Build GRCh38
rs769144942 Chr.10: 97589995-97589995 on Build GRCh38
rs769018488 Chr.10: 97589866-97589866 on Build GRCh38
rs747477606 Chr.10: 97589864-97589864 on Build GRCh38
rs778440495 Chr.10: 97589800-97589800 on Build GRCh38
rs747764983 Chr.10: 97589813-97589813 on Build GRCh38
rs369324110 Chr.10: 97589854-97589854 on Build GRCh38
rs565197999 Chr.10: 97589853-97589853 on Build GRCh38
rs933387763 Chr.10: 97589939-97589939 on Build GRCh38
rs1048754727 Chr.10: 97589789-97589789 on Build GRCh38
rs751546544 Chr.10: 97589923-97589923 on Build GRCh38
rs374772045 Chr.10: 97589847-97589847 on Build GRCh38
rs766524986 Chr.10: 97589921-97589921 on Build GRCh38
rs773212270 Chr.10: 97589914-97589914 on Build GRCh38
rs1048628524 Chr.10: 97589799-97589799 on Build GRCh38
rs756793346 Chr.10: 97589978-97589978 on Build GRCh38
rs560755279 Chr.10: 97589780-97589780 on Build GRCh38
rs763146588 Chr.10: 97589915-97589915 on Build GRCh38
rs911146343 Chr.10: 97589814-97589814 on Build GRCh38
rs777038816 Chr.10: 97589872-97589872 on Build GRCh38
rs762003725 Chr.10: 97589894-97589894 on Build GRCh38
rs752532956 Chr.10: 97589960-97589960 on Build GRCh38
rs747569609 Chr.10: 97589990-97589990 on Build GRCh38

View Full Product Details