Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95448302-95448527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM81B
Entrez Gene ID
153643
Gene Name
family with sequence similarity 81 member B
Gene Aliases
-
UniGene
Hs.276287
Gene Chromosome Location
Chr.5: 95384793-95450441 on Build GRCh38
Transcripts
NM_152548.2 XM_005271905.2 XM_005271906.4 XM_011543209.1 XM_011543210.2 XM_017009118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391396 c.1102C>G p.Q368E Chr.5: 95448341-95448341 on Build GRCh38
6047376 c.1095T>C p.I365I Chr.5: 95448334-95448334 on Build GRCh38
7197200 c.1064T>A p.L355Q Chr.5: 95448303-95448303 on Build GRCh38
1206452 c.1164G>A p.M388I Chr.5: 95448403-95448403 on Build GRCh38
7281914 c.1185C>T p.I395I Chr.5: 95448424-95448424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767517546 Chr.5: 95448483-95448483 on Build GRCh38
rs199697879 Chr.5: 95448429-95448429 on Build GRCh38
rs762124160 Chr.5: 95448442-95448442 on Build GRCh38
rs148203459 Chr.5: 95448378-95448378 on Build GRCh38
rs756787447 Chr.5: 95448341-95448341 on Build GRCh38
rs749381783 Chr.5: 95448421-95448421 on Build GRCh38
rs972991088 Chr.5: 95448435-95448435 on Build GRCh38
rs775481994 Chr.5: 95448399-95448399 on Build GRCh38
rs371659677 Chr.5: 95448345-95448345 on Build GRCh38
rs200218795 Chr.5: 95448374-95448374 on Build GRCh38
rs1052638746 Chr.5: 95448426-95448426 on Build GRCh38
rs767951439 Chr.5: 95448454-95448454 on Build GRCh38
rs746265983 Chr.5: 95448361-95448365 on Build GRCh38
rs947043521 Chr.5: 95448339-95448339 on Build GRCh38
rs761205189 Chr.5: 95448488-95448488 on Build GRCh38
rs532902276 Chr.5: 95448483-95448483 on Build GRCh38
rs147322096 Chr.5: 95448379-95448379 on Build GRCh38
rs1017163410 Chr.5: 95448402-95448402 on Build GRCh38
rs1003121638 Chr.5: 95448326-95448326 on Build GRCh38
rs768820861 Chr.5: 95448423-95448423 on Build GRCh38
rs772473395 Chr.5: 95448436-95448437 on Build GRCh38
rs77360423 Chr.5: 95448422-95448422 on Build GRCh38

View Full Product Details