Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108628088-108628323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102B
Entrez Gene ID
284611
Gene Name
family with sequence similarity 102 member B
Gene Aliases
SYM-3B
UniGene
Hs.200230
Gene Chromosome Location
Chr.1: 108560349-108639327 on Build GRCh38
Transcripts
NM_001010883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5936872 c.578C>T p.S193F Chr.1: 108628188-108628188 on Build GRCh38
4410149 c.593T>G p.L198R Chr.1: 108628203-108628203 on Build GRCh38
236096 c.466C>A p.L156I Chr.1: 108628169-108628169 on Build GRCh38
1243236 c.588C>T p.D196D Chr.1: 108628198-108628198 on Build GRCh38
5936873 c.485C>T p.S162F Chr.1: 108628188-108628188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775988340 Chr.1: 108628239-108628239 on Build GRCh38
rs777038027 Chr.1: 108628209-108628209 on Build GRCh38
rs762625248 Chr.1: 108628118-108628118 on Build GRCh38
rs144437284 Chr.1: 108628249-108628249 on Build GRCh38
rs371756924 Chr.1: 108628198-108628198 on Build GRCh38
rs186994288 Chr.1: 108628281-108628281 on Build GRCh38
rs755924809 Chr.1: 108628149-108628149 on Build GRCh38
rs1010689388 Chr.1: 108628158-108628158 on Build GRCh38
rs1020870317 Chr.1: 108628169-108628169 on Build GRCh38
rs373686490 Chr.1: 108628268-108628268 on Build GRCh38
rs954787903 Chr.1: 108628186-108628186 on Build GRCh38
rs182933966 Chr.1: 108628119-108628119 on Build GRCh38
rs1005083236 Chr.1: 108628221-108628221 on Build GRCh38
rs567848198 Chr.1: 108628263-108628263 on Build GRCh38
rs760059745 Chr.1: 108628212-108628212 on Build GRCh38
rs147603320 Chr.1: 108628228-108628228 on Build GRCh38
rs2133198 Chr.1: 108628124-108628124 on Build GRCh38
rs771513451 Chr.1: 108628199-108628199 on Build GRCh38
rs779891677 Chr.1: 108628288-108628288 on Build GRCh38
rs113778227 Chr.1: 108628120-108628120 on Build GRCh38
rs865775446 Chr.1: 108628188-108628188 on Build GRCh38
rs780960396 Chr.1: 108628145-108628145 on Build GRCh38
rs746304135 Chr.1: 108628243-108628243 on Build GRCh38
rs779768955 Chr.1: 108628152-108628152 on Build GRCh38
rs577018597 Chr.1: 108628287-108628287 on Build GRCh38
rs111566057 Chr.1: 108628151-108628151 on Build GRCh38
rs770285331 Chr.1: 108628229-108628229 on Build GRCh38
rs376063075 Chr.1: 108628146-108628146 on Build GRCh38
rs761257890 Chr.1: 108628270-108628270 on Build GRCh38
rs748376815 Chr.1: 108628161-108628161 on Build GRCh38
rs533533263 Chr.1: 108628280-108628280 on Build GRCh38
rs772277143 Chr.1: 108628193-108628193 on Build GRCh38
rs147413102 Chr.1: 108628294-108628294 on Build GRCh38
rs777968745 Chr.1: 108628195-108628195 on Build GRCh38

View Full Product Details