Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87792608-87792851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
UniGene
Hs.435948
Gene Chromosome Location
Chr.10: 87751512-87841345 on Build GRCh38
Transcripts
NM_001321967.1 NM_001321968.1 NM_001321969.1 NM_032810.3 NR_135914.1 XM_005270252.4 XM_011540302.1 XM_011540303.2 XM_017016847.1 XM_017016848.1 XM_017016849.1 XM_017016850.1 XM_017016851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347520 c.208T>G p.S70A Chr.10: 87792710-87792710 on Build GRCh38
1349455 c.188G>A p.G63E Chr.10: 87792730-87792730 on Build GRCh38
1171729 c.217G>C p.E73Q Chr.10: 87792701-87792701 on Build GRCh38
4663927 c.164_166delCAG p.A55delA Chr.10: 87792752-87792754 on Build GRCh38
6518090 c.183A>T p.Q61H Chr.10: 87792735-87792735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944940345 Chr.10: 87792639-87792639 on Build GRCh38
rs757087401 Chr.10: 87792719-87792719 on Build GRCh38
rs1050904658 Chr.10: 87792762-87792762 on Build GRCh38
rs899530214 Chr.10: 87792714-87792714 on Build GRCh38
rs760173843 Chr.10: 87792769-87792772 on Build GRCh38
rs371862255 Chr.10: 87792743-87792743 on Build GRCh38
rs1048640955 Chr.10: 87792811-87792811 on Build GRCh38
rs756304470 Chr.10: 87792684-87792684 on Build GRCh38
rs530630863 Chr.10: 87792671-87792671 on Build GRCh38
rs778119962 Chr.10: 87792694-87792694 on Build GRCh38
rs749081136 Chr.10: 87792702-87792702 on Build GRCh38
rs921124074 Chr.10: 87792631-87792631 on Build GRCh38
rs774873129 Chr.10: 87792781-87792781 on Build GRCh38
rs375208407 Chr.10: 87792763-87792763 on Build GRCh38
rs766871296 Chr.10: 87792807-87792807 on Build GRCh38
rs563335586 Chr.10: 87792632-87792632 on Build GRCh38
rs996594836 Chr.10: 87792738-87792738 on Build GRCh38
rs776462080 Chr.10: 87792801-87792801 on Build GRCh38
rs755061549 Chr.10: 87792645-87792645 on Build GRCh38
rs912112877 Chr.10: 87792629-87792629 on Build GRCh38
rs771540170 Chr.10: 87792774-87792774 on Build GRCh38
rs768189320 Chr.10: 87792795-87792795 on Build GRCh38
rs778802074 Chr.10: 87792732-87792732 on Build GRCh38
rs767526639 Chr.10: 87792658-87792658 on Build GRCh38
rs746583699 Chr.10: 87792784-87792784 on Build GRCh38
rs543985404 Chr.10: 87792812-87792812 on Build GRCh38
rs763385233 Chr.10: 87792804-87792804 on Build GRCh38

View Full Product Details