Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2232147-2232372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E4F1
Entrez Gene ID
1877
Gene Name
E4F transcription factor 1
Gene Aliases
E4F
UniGene
Hs.513268
Gene Chromosome Location
Chr.16: 2223488-2235742 on Build GRCh38
Transcripts
NM_001288776.1 NM_001288778.1 NM_004424.4 XM_005255155.1 XM_005255156.1 XM_006720858.1 XM_011522402.1 XM_011522403.1 XM_017023010.1 XM_017023011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
237328 c.609+7C>T p.? Chr.16: 2232371-2232371 on Build GRCh38
6651084 c.570C>T p.G190G Chr.16: 2232325-2232325 on Build GRCh38
968657 c.564G>T p.K188N Chr.16: 2232319-2232319 on Build GRCh38
3667790 c.568G>A p.G190S Chr.16: 2232323-2232323 on Build GRCh38
5462515 c.590G>A p.C197Y Chr.16: 2232345-2232345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753059691 Chr.16: 2232298-2232298 on Build GRCh38
rs758627188 Chr.16: 2232235-2232235 on Build GRCh38
rs751937000 Chr.16: 2232236-2232236 on Build GRCh38
rs973510016 Chr.16: 2232169-2232169 on Build GRCh38
rs770485404 Chr.16: 2232193-2232193 on Build GRCh38
rs761799045 Chr.16: 2232263-2232263 on Build GRCh38
rs142656555 Chr.16: 2232243-2232243 on Build GRCh38
rs771680213 Chr.16: 2232244-2232244 on Build GRCh38
rs767987855 Chr.16: 2232297-2232297 on Build GRCh38
rs750488930 Chr.16: 2232270-2232270 on Build GRCh38
rs1039633956 Chr.16: 2232247-2232247 on Build GRCh38
rs748690987 Chr.16: 2232242-2232242 on Build GRCh38
rs576266110 Chr.16: 2232275-2232275 on Build GRCh38
rs776357419 Chr.16: 2232257-2232257 on Build GRCh38
rs765357279 Chr.16: 2232221-2232221 on Build GRCh38
rs375178489 Chr.16: 2232223-2232223 on Build GRCh38
rs765140149 Chr.16: 2232264-2232264 on Build GRCh38
rs568521071 Chr.16: 2232210-2232210 on Build GRCh38
rs769735039 Chr.16: 2232340-2232340 on Build GRCh38
rs757917390 Chr.16: 2232307-2232307 on Build GRCh38
rs755324797 Chr.16: 2232287-2232287 on Build GRCh38
rs781739631 Chr.16: 2232241-2232241 on Build GRCh38
rs779615574 Chr.16: 2232318-2232318 on Build GRCh38
rs149258624 Chr.16: 2232274-2232274 on Build GRCh38
rs768357528 Chr.16: 2232321-2232321 on Build GRCh38
rs776466711 Chr.16: 2232331-2232331 on Build GRCh38
rs26839 Chr.16: 2232255-2232255 on Build GRCh38
rs747936086 Chr.16: 2232339-2232339 on Build GRCh38
rs773805400 Chr.16: 2232196-2232196 on Build GRCh38
rs755412650 Chr.16: 2232240-2232240 on Build GRCh38
rs775154393 Chr.16: 2232250-2232250 on Build GRCh38
rs763064601 Chr.16: 2232349-2232349 on Build GRCh38
rs760502409 Chr.16: 2232211-2232211 on Build GRCh38
rs143442513 Chr.16: 2232197-2232197 on Build GRCh38
rs746661970 Chr.16: 2232320-2232320 on Build GRCh38
rs138184443 Chr.16: 2232186-2232186 on Build GRCh38
rs921147890 Chr.16: 2232254-2232254 on Build GRCh38
rs756620926 Chr.16: 2232304-2232304 on Build GRCh38
rs945863105 Chr.16: 2232354-2232354 on Build GRCh38
rs778183040 Chr.16: 2232305-2232305 on Build GRCh38
rs773050360 Chr.16: 2232346-2232346 on Build GRCh38
rs748694000 Chr.16: 2232185-2232185 on Build GRCh38
rs879213356 Chr.16: 2232217-2232217 on Build GRCh38
rs753827991 Chr.16: 2232216-2232216 on Build GRCh38

View Full Product Details