Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34292289-34292558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCY
Entrez Gene ID
191
Gene Name
adenosylhomocysteinase
Gene Aliases
SAHH, adoHcyase
UniGene
Hs.388004
Gene Chromosome Location
Chr.20: 34235012-34311976 on Build GRCh38
Transcripts
NM_000687.3 NM_001161766.1 NM_001322084.1 NM_001322085.1 NM_001322086.1 XM_005260317.2 XM_011528656.2 XM_011528657.2 XM_011528658.2 XM_011528659.1 XM_017027708.1 XM_017027709.1 XM_017027710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6417942 c.346C>T p.Q116* Chr.20: 34292457-34292457 on Build GRCh38
5857714 c.388G>A p.D130N Chr.20: 34292415-34292415 on Build GRCh38
127278 c.399C>T p.G133G Chr.20: 34292404-34292404 on Build GRCh38
4833997 c.408C>T p.T136T Chr.20: 34292395-34292395 on Build GRCh38
1713377 c.407C>T p.T136I Chr.20: 34292396-34292396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11552698 Chr.20: 34292374-34292374 on Build GRCh38
rs141935523 Chr.20: 34292437-34292437 on Build GRCh38
rs750090362 Chr.20: 34292383-34292383 on Build GRCh38
rs149484182 Chr.20: 34292404-34292404 on Build GRCh38
rs746290790 Chr.20: 34292528-34292528 on Build GRCh38
rs779559117 Chr.20: 34292323-34292323 on Build GRCh38
rs11552700 Chr.20: 34292400-34292400 on Build GRCh38
rs752121128 Chr.20: 34292500-34292500 on Build GRCh38
rs748608602 Chr.20: 34292324-34292324 on Build GRCh38
rs778383891 Chr.20: 34292409-34292409 on Build GRCh38
rs754422062 Chr.20: 34292407-34292407 on Build GRCh38
rs757079296 Chr.20: 34292380-34292380 on Build GRCh38
rs59620110 Chr.20: 34292530-34292530 on Build GRCh38
rs745484768 Chr.20: 34292313-34292313 on Build GRCh38
rs1021875336 Chr.20: 34292403-34292403 on Build GRCh38
rs773578300 Chr.20: 34292338-34292338 on Build GRCh38
rs781639233 Chr.20: 34292419-34292419 on Build GRCh38
rs760451324 Chr.20: 34292482-34292482 on Build GRCh38
rs772704608 Chr.20: 34292325-34292325 on Build GRCh38
rs778704134 Chr.20: 34292522-34292523 on Build GRCh38
rs121918607 Chr.20: 34292467-34292467 on Build GRCh38
rs553442034 Chr.20: 34292410-34292410 on Build GRCh38
rs755697459 Chr.20: 34292387-34292387 on Build GRCh38
rs121918608 Chr.20: 34292375-34292375 on Build GRCh38
rs746089763 Chr.20: 34292424-34292424 on Build GRCh38
rs768582053 Chr.20: 34292445-34292445 on Build GRCh38
rs201902693 Chr.20: 34292494-34292494 on Build GRCh38
rs765416734 Chr.20: 34292354-34292354 on Build GRCh38
rs761681773 Chr.20: 34292452-34292452 on Build GRCh38
rs926001235 Chr.20: 34292479-34292479 on Build GRCh38
rs199616559 Chr.20: 34292398-34292398 on Build GRCh38
rs778722859 Chr.20: 34292451-34292451 on Build GRCh38
rs747915996 Chr.20: 34292473-34292473 on Build GRCh38
rs558714552 Chr.20: 34292339-34292339 on Build GRCh38
rs766103083 Chr.20: 34292485-34292485 on Build GRCh38
rs143931269 Chr.20: 34292413-34292413 on Build GRCh38
rs370629168 Chr.20: 34292502-34292502 on Build GRCh38
rs375253690 Chr.20: 34292347-34292347 on Build GRCh38
rs556286156 Chr.20: 34292372-34292372 on Build GRCh38
rs753399141 Chr.20: 34292491-34292491 on Build GRCh38
rs759780277 Chr.20: 34292351-34292351 on Build GRCh38
rs910267027 Chr.20: 34292486-34292486 on Build GRCh38
rs41301825 Chr.20: 34292436-34292436 on Build GRCh38
rs11552699 Chr.20: 34292379-34292379 on Build GRCh38
rs978859696 Chr.20: 34292457-34292457 on Build GRCh38
rs11552696 Chr.20: 34292363-34292363 on Build GRCh38
rs780867268 Chr.20: 34292308-34292308 on Build GRCh38
rs577419934 Chr.20: 34292381-34292381 on Build GRCh38
rs11552697 Chr.20: 34292428-34292428 on Build GRCh38
rs767304411 Chr.20: 34292474-34292474 on Build GRCh38
rs146167334 Chr.20: 34292475-34292475 on Build GRCh38
rs781762671 Chr.20: 34292517-34292517 on Build GRCh38
rs747425459 Chr.20: 34292346-34292346 on Build GRCh38

View Full Product Details