Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27292202-27292463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EQTN
Entrez Gene ID
54586
Gene Name
equatorin
Gene Aliases
AFAF, C9orf11, SPACA8
UniGene
Hs.163070
Gene Chromosome Location
Chr.9: 27284654-27297139 on Build GRCh38
Transcripts
NM_001161585.1 NM_020641.2 XM_011517920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4449753 c.316delG p.E106fs*33 Chr.9: 27292461-27292461 on Build GRCh38
4543267 c.334G>A p.E112K Chr.9: 27292443-27292443 on Build GRCh38
7105851 c.321_322insAA p.S108fs*32 Chr.9: 27292455-27292456 on Build GRCh38
2771644 c.352G>A p.E118K Chr.9: 27292425-27292425 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376999420 Chr.9: 27292355-27292355 on Build GRCh38
rs760832540 Chr.9: 27292420-27292420 on Build GRCh38
rs764664576 Chr.9: 27292337-27292337 on Build GRCh38
rs780534327 Chr.9: 27292352-27292352 on Build GRCh38
rs752563294 Chr.9: 27292430-27292430 on Build GRCh38
rs748237320 Chr.9: 27292279-27292279 on Build GRCh38
rs754231767 Chr.9: 27292420-27292420 on Build GRCh38
rs988030786 Chr.9: 27292271-27292271 on Build GRCh38
rs766619273 Chr.9: 27292421-27292421 on Build GRCh38
rs3739534 Chr.9: 27292281-27292281 on Build GRCh38
rs768952797 Chr.9: 27292360-27292360 on Build GRCh38
rs746510090 Chr.9: 27292348-27292348 on Build GRCh38
rs909510500 Chr.9: 27292303-27292303 on Build GRCh38
rs772314637 Chr.9: 27292405-27292405 on Build GRCh38
rs777928503 Chr.9: 27292314-27292314 on Build GRCh38
rs764904472 Chr.9: 27292426-27292426 on Build GRCh38
rs770823243 Chr.9: 27292307-27292307 on Build GRCh38
rs147923701 Chr.9: 27292363-27292364 on Build GRCh38
rs73643174 Chr.9: 27292287-27292287 on Build GRCh38
rs966114311 Chr.9: 27292237-27292237 on Build GRCh38
rs748267115 Chr.9: 27292381-27292381 on Build GRCh38
rs759548451 Chr.9: 27292425-27292425 on Build GRCh38
rs535329909 Chr.9: 27292322-27292322 on Build GRCh38
rs773345459 Chr.9: 27292417-27292417 on Build GRCh38
rs774784847 Chr.9: 27292372-27292372 on Build GRCh38
rs917035118 Chr.9: 27292268-27292268 on Build GRCh38
rs10757650 Chr.9: 27292236-27292236 on Build GRCh38
rs776649941 Chr.9: 27292423-27292423 on Build GRCh38

View Full Product Details