Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137914617-137914870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1B
Entrez Gene ID
774
Gene Name
calcium voltage-gated channel subunit alpha1 B
Gene Aliases
BIII, CACNL1A5, CACNN, Cav2.2, DYT23
UniGene
Hs.495522
Gene Chromosome Location
Chr.9: 137877789-138124624 on Build GRCh38
Transcripts
NM_000718.3 NM_001243812.1 XM_011518991.2 XM_011518992.2 XM_011518993.2 XM_011518994.2 XM_017015115.1 XM_017015116.1 XM_017015117.1 XM_017015118.1 XM_017015119.1 XM_017015120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6115309 c.634G>T p.V212L Chr.9: 137914665-137914665 on Build GRCh38
7303474 c.655G>T p.A219S Chr.9: 137914686-137914686 on Build GRCh38
753007 c.717C>T p.I239I Chr.9: 137914748-137914748 on Build GRCh38
4393766 c.729G>A p.E243E Chr.9: 137914760-137914760 on Build GRCh38
7196996 c.713C>A p.A238D Chr.9: 137914744-137914744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200521478 Chr.9: 137914839-137914839 on Build GRCh38
rs773179550 Chr.9: 137914736-137914736 on Build GRCh38
rs199792680 Chr.9: 137914834-137914834 on Build GRCh38
rs763939363 Chr.9: 137914771-137914771 on Build GRCh38
rs769630341 Chr.9: 137914680-137914680 on Build GRCh38
rs751021410 Chr.9: 137914650-137914650 on Build GRCh38
rs761312939 Chr.9: 137914835-137914835 on Build GRCh38
rs372939958 Chr.9: 137914641-137914641 on Build GRCh38
rs760088580 Chr.9: 137914815-137914815 on Build GRCh38
rs11137301 Chr.9: 137914847-137914847 on Build GRCh38
rs757142084 Chr.9: 137914667-137914667 on Build GRCh38
rs200274455 Chr.9: 137914746-137914746 on Build GRCh38
rs779795778 Chr.9: 137914693-137914693 on Build GRCh38
rs997663132 Chr.9: 137914777-137914777 on Build GRCh38
rs760741224 Chr.9: 137914739-137914739 on Build GRCh38
rs532639291 Chr.9: 137914846-137914846 on Build GRCh38
rs781168541 Chr.9: 137914670-137914670 on Build GRCh38
rs150072064 Chr.9: 137914816-137914816 on Build GRCh38
rs567155794 Chr.9: 137914819-137914819 on Build GRCh38
rs1039293406 Chr.9: 137914643-137914643 on Build GRCh38
rs201810627 Chr.9: 137914708-137914708 on Build GRCh38
rs772290130 Chr.9: 137914733-137914733 on Build GRCh38
rs777326436 Chr.9: 137914799-137914799 on Build GRCh38
rs958126621 Chr.9: 137914823-137914823 on Build GRCh38
rs748352783 Chr.9: 137914715-137914715 on Build GRCh38
rs762523004 Chr.9: 137914820-137914820 on Build GRCh38
rs745795777 Chr.9: 137914679-137914679 on Build GRCh38
rs771131603 Chr.9: 137914785-137914785 on Build GRCh38

View Full Product Details