Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31535653-31535882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf22
Entrez Gene ID
55322
Gene Name
chromosome 5 open reading frame 22
Gene Aliases
-
UniGene
Hs.519246
Gene Chromosome Location
Chr.5: 31532086-31555058 on Build GRCh38
Transcripts
NM_018356.2 NR_134298.1 XM_006714479.2 XM_006714480.3 XM_011514062.2 XM_017009607.1 XM_017009608.1 XM_017009609.1 XM_017009610.1 XM_017009611.1 XM_017009612.1 XM_017009613.1 XM_017009614.1 XM_017009615.1 XM_017009616.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1311091 c.323G>C p.R108T Chr.5: 31535839-31535839 on Build GRCh38
5975576 c.325G>A p.E109K Chr.5: 31535841-31535841 on Build GRCh38
4543195 c.332G>C p.R111T Chr.5: 31535848-31535848 on Build GRCh38
137888 c.228-1G>A p.? Chr.5: 31535743-31535743 on Build GRCh38
266143 c.289A>G p.I97V Chr.5: 31535805-31535805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765195777 Chr.5: 31535729-31535729 on Build GRCh38
rs200760829 Chr.5: 31535827-31535827 on Build GRCh38
rs780737764 Chr.5: 31535805-31535805 on Build GRCh38
rs758261011 Chr.5: 31535762-31535762 on Build GRCh38
rs1003179077 Chr.5: 31535775-31535775 on Build GRCh38
rs777201268 Chr.5: 31535822-31535822 on Build GRCh38
rs376281573 Chr.5: 31535845-31535845 on Build GRCh38
rs567965415 Chr.5: 31535826-31535826 on Build GRCh38
rs764325242 Chr.5: 31535718-31535718 on Build GRCh38
rs147175944 Chr.5: 31535808-31535808 on Build GRCh38
rs994947722 Chr.5: 31535692-31535692 on Build GRCh38
rs776735936 Chr.5: 31535726-31535726 on Build GRCh38
rs200288266 Chr.5: 31535800-31535800 on Build GRCh38
rs372673202 Chr.5: 31535728-31535728 on Build GRCh38
rs773127089 Chr.5: 31535791-31535792 on Build GRCh38
rs372061653 Chr.5: 31535752-31535752 on Build GRCh38
rs772582721 Chr.5: 31535830-31535830 on Build GRCh38
rs766207392 Chr.5: 31535776-31535776 on Build GRCh38
rs747409418 Chr.5: 31535840-31535840 on Build GRCh38
rs751411764 Chr.5: 31535779-31535779 on Build GRCh38
rs905425410 Chr.5: 31535745-31535745 on Build GRCh38
rs760831459 Chr.5: 31535703-31535703 on Build GRCh38
rs748803553 Chr.5: 31535848-31535848 on Build GRCh38
rs754627032 Chr.5: 31535801-31535801 on Build GRCh38
rs755819547 Chr.5: 31535799-31535799 on Build GRCh38
rs765213484 Chr.5: 31535748-31535749 on Build GRCh38
rs755637447 Chr.5: 31535818-31535818 on Build GRCh38
rs897200302 Chr.5: 31535727-31535727 on Build GRCh38
rs775786385 Chr.5: 31535838-31535838 on Build GRCh38

View Full Product Details