Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89100690-89100939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF3
Entrez Gene ID
197322
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
UniGene
Hs.461727
Gene Chromosome Location
Chr.16: 89093809-89160556 on Build GRCh38
Transcripts
NM_001127214.3 NM_001243279.2 NM_001284316.1 NM_174917.4 NR_104293.1 XM_005256293.2 XM_011522942.1 XM_011522943.1 XM_017023018.1 XM_017023019.1 XM_017023020.1 XM_017023021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3378025 c.189C>T p.H63H Chr.16: 89100870-89100870 on Build GRCh38
308835 c.194G>A p.R65H Chr.16: 89100875-89100875 on Build GRCh38
3513246 c.80G>A p.R27K Chr.16: 89100761-89100761 on Build GRCh38
6592540 c.148G>T p.A50S Chr.16: 89100829-89100829 on Build GRCh38
5516064 c.231C>T p.R77R Chr.16: 89100912-89100912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931176162 Chr.16: 89100756-89100756 on Build GRCh38
rs779775679 Chr.16: 89100796-89100796 on Build GRCh38
rs770572252 Chr.16: 89100873-89100873 on Build GRCh38
rs745684193 Chr.16: 89100833-89100839 on Build GRCh38
rs147044602 Chr.16: 89100852-89100852 on Build GRCh38
rs767348758 Chr.16: 89100815-89100815 on Build GRCh38
rs751461445 Chr.16: 89100747-89100747 on Build GRCh38
rs562260243 Chr.16: 89100870-89100870 on Build GRCh38
rs375374971 Chr.16: 89100724-89100724 on Build GRCh38
rs369499144 Chr.16: 89100827-89100827 on Build GRCh38
rs763197756 Chr.16: 89100906-89100906 on Build GRCh38
rs1040960753 Chr.16: 89100711-89100711 on Build GRCh38
rs775259772 Chr.16: 89100889-89100889 on Build GRCh38
rs754161182 Chr.16: 89100767-89100767 on Build GRCh38
rs746564011 Chr.16: 89100837-89100837 on Build GRCh38
rs572357895 Chr.16: 89100746-89100746 on Build GRCh38
rs779450250 Chr.16: 89100786-89100786 on Build GRCh38
rs746400895 Chr.16: 89100731-89100731 on Build GRCh38
rs373274928 Chr.16: 89100839-89100839 on Build GRCh38
rs141518662 Chr.16: 89100826-89100826 on Build GRCh38
rs147915828 Chr.16: 89100810-89100810 on Build GRCh38
rs757664305 Chr.16: 89100780-89100780 on Build GRCh38
rs1028917212 Chr.16: 89100814-89100814 on Build GRCh38
rs11547019 Chr.16: 89100730-89100730 on Build GRCh38
rs745633046 Chr.16: 89100875-89100875 on Build GRCh38
rs767573178 Chr.16: 89100750-89100750 on Build GRCh38
rs774117419 Chr.16: 89100874-89100874 on Build GRCh38
rs748883934 Chr.16: 89100872-89100872 on Build GRCh38
rs752720474 Chr.16: 89100753-89100753 on Build GRCh38
rs747598756 Chr.16: 89100800-89100800 on Build GRCh38
rs746334624 Chr.16: 89100789-89100789 on Build GRCh38
rs901081235 Chr.16: 89100712-89100712 on Build GRCh38
rs7201122 Chr.16: 89100732-89100732 on Build GRCh38
rs756300619 Chr.16: 89100723-89100723 on Build GRCh38
rs753339356 Chr.16: 89100920-89100920 on Build GRCh38
rs144711526 Chr.16: 89100797-89100797 on Build GRCh38
rs755491777 Chr.16: 89100917-89100917 on Build GRCh38
rs1035298569 Chr.16: 89100752-89100752 on Build GRCh38
rs764393364 Chr.16: 89100764-89100764 on Build GRCh38
rs762719449 Chr.16: 89100740-89100740 on Build GRCh38
rs564576843 Chr.16: 89100805-89100805 on Build GRCh38
rs766802453 Chr.16: 89100908-89100908 on Build GRCh38
rs759448696 Chr.16: 89100811-89100811 on Build GRCh38
rs202121474 Chr.16: 89100864-89100864 on Build GRCh38
rs771946908 Chr.16: 89100881-89100881 on Build GRCh38
rs552894451 Chr.16: 89100737-89100737 on Build GRCh38
rs534594073 Chr.16: 89100734-89100734 on Build GRCh38
rs780402535 Chr.16: 89100860-89100860 on Build GRCh38
rs901883302 Chr.16: 89100804-89100804 on Build GRCh38
rs765633249 Chr.16: 89100840-89100840 on Build GRCh38
rs999563327 Chr.16: 89100719-89100719 on Build GRCh38
rs771481057 Chr.16: 89100726-89100726 on Build GRCh38
rs200536797 Chr.16: 89100871-89100871 on Build GRCh38
rs202182978 Chr.16: 89100709-89100709 on Build GRCh38
rs781163359 Chr.16: 89100713-89100713 on Build GRCh38
rs922581606 Chr.16: 89100841-89100841 on Build GRCh38
rs751551226 Chr.16: 89100710-89100710 on Build GRCh38
rs34972688 Chr.16: 89100801-89100801 on Build GRCh38
rs762053293 Chr.16: 89100899-89100899 on Build GRCh38
rs752913078 Chr.16: 89100718-89100718 on Build GRCh38
rs545886514 Chr.16: 89100803-89100803 on Build GRCh38
rs138156311 Chr.16: 89100919-89100919 on Build GRCh38
rs749661738 Chr.16: 89100725-89100725 on Build GRCh38
rs529560098 Chr.16: 89100892-89100892 on Build GRCh38
rs776862565 Chr.16: 89100898-89100898 on Build GRCh38
rs750707295 Chr.16: 89100902-89100902 on Build GRCh38
rs772907663 Chr.16: 89100739-89100739 on Build GRCh38
rs746501634 Chr.16: 89100911-89100911 on Build GRCh38
rs758856054 Chr.16: 89100853-89100853 on Build GRCh38
rs775551806 Chr.16: 89100816-89100816 on Build GRCh38
rs549664473 Chr.16: 89100791-89100791 on Build GRCh38
rs756279730 Chr.16: 89100761-89100761 on Build GRCh38

View Full Product Details