Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 24678544-24678813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAP1
Entrez Gene ID
23742
Gene Name
nuclear pore associated protein 1
Gene Aliases
C15orf2
UniGene
Hs.649663
Gene Chromosome Location
Chr.15: 24675394-24683446 on Build GRCh38
Transcripts
NM_018958.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3500137 c.2681G>A p.G894E Chr.15: 24678548-24678548 on Build GRCh38
7422229 c.2940T>A p.S980S Chr.15: 24678807-24678807 on Build GRCh38
3690315 c.2810G>A p.S937N Chr.15: 24678677-24678677 on Build GRCh38
3500139 c.2717G>A p.G906E Chr.15: 24678584-24678584 on Build GRCh38
6521888 c.2738C>A p.S913Y Chr.15: 24678605-24678605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201694207 Chr.15: 24678716-24678716 on Build GRCh38
rs199762732 Chr.15: 24678640-24678640 on Build GRCh38
rs754197057 Chr.15: 24678594-24678594 on Build GRCh38
rs766971774 Chr.15: 24678586-24678586 on Build GRCh38
rs368505553 Chr.15: 24678584-24678584 on Build GRCh38
rs757277275 Chr.15: 24678762-24678762 on Build GRCh38
rs765574312 Chr.15: 24678715-24678715 on Build GRCh38
rs758481864 Chr.15: 24678622-24678622 on Build GRCh38
rs539929155 Chr.15: 24678761-24678761 on Build GRCh38
rs770892959 Chr.15: 24678642-24678642 on Build GRCh38
rs758664880 Chr.15: 24678757-24678757 on Build GRCh38
rs754325256 Chr.15: 24678695-24678695 on Build GRCh38
rs747393900 Chr.15: 24678635-24678635 on Build GRCh38
rs371377318 Chr.15: 24678789-24678789 on Build GRCh38
rs763776658 Chr.15: 24678732-24678732 on Build GRCh38
rs763630353 Chr.15: 24678569-24678569 on Build GRCh38
rs867699058 Chr.15: 24678680-24678680 on Build GRCh38
rs140150292 Chr.15: 24678774-24678774 on Build GRCh38
rs375207590 Chr.15: 24678718-24678718 on Build GRCh38
rs759835421 Chr.15: 24678712-24678712 on Build GRCh38
rs372965255 Chr.15: 24678600-24678600 on Build GRCh38
rs868570818 Chr.15: 24678718-24678719 on Build GRCh38
rs761376976 Chr.15: 24678677-24678677 on Build GRCh38
rs113278134 Chr.15: 24678686-24678686 on Build GRCh38
rs752495951 Chr.15: 24678656-24678658 on Build GRCh38
rs769572754 Chr.15: 24678657-24678657 on Build GRCh38
rs375473121 Chr.15: 24678610-24678610 on Build GRCh38
rs775010025 Chr.15: 24678664-24678664 on Build GRCh38
rs1001161563 Chr.15: 24678722-24678722 on Build GRCh38
rs372009647 Chr.15: 24678674-24678674 on Build GRCh38
rs149131090 Chr.15: 24678648-24678648 on Build GRCh38
rs267604139 Chr.15: 24678779-24678779 on Build GRCh38
rs751111039 Chr.15: 24678577-24678577 on Build GRCh38
rs749060463 Chr.15: 24678788-24678788 on Build GRCh38
rs369566519 Chr.15: 24678759-24678759 on Build GRCh38
rs151169751 Chr.15: 24678685-24678685 on Build GRCh38
rs769614427 Chr.15: 24678773-24678773 on Build GRCh38
rs767423190 Chr.15: 24678571-24678573 on Build GRCh38
rs368406423 Chr.15: 24678666-24678666 on Build GRCh38
rs753004382 Chr.15: 24678727-24678727 on Build GRCh38
rs781003390 Chr.15: 24678763-24678763 on Build GRCh38
rs938040169 Chr.15: 24678576-24678576 on Build GRCh38
rs745750086 Chr.15: 24678772-24678772 on Build GRCh38
rs955882642 Chr.15: 24678660-24678660 on Build GRCh38
rs774096468 Chr.15: 24678790-24678790 on Build GRCh38
rs755741411 Chr.15: 24678719-24678720 on Build GRCh38
rs547257889 Chr.15: 24678605-24678605 on Build GRCh38
rs768411016 Chr.15: 24678671-24678671 on Build GRCh38
rs762382044 Chr.15: 24678567-24678567 on Build GRCh38
rs34413216 Chr.15: 24678652-24678652 on Build GRCh38

View Full Product Details