Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50535212-50535473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAF1
Entrez Gene ID
11124
Gene Name
Fas associated factor 1
Gene Aliases
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1
UniGene
Hs.530402
Gene Chromosome Location
Chr.1: 50441263-50960264 on Build GRCh38
Transcripts
NM_007051.2 XM_017000136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6126998 c.1415C>T p.T472I Chr.1: 50535448-50535448 on Build GRCh38
3400863 c.1406-2A>G p.? Chr.1: 50535459-50535459 on Build GRCh38
1738559 c.1420G>A p.D474N Chr.1: 50535443-50535443 on Build GRCh38
2192228 c.1432A>G p.M478V Chr.1: 50535431-50535431 on Build GRCh38
6126999 c.689C>T p.T230I Chr.1: 50535448-50535448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781750005 Chr.1: 50535360-50535360 on Build GRCh38
rs1021736013 Chr.1: 50535268-50535268 on Build GRCh38
rs752894904 Chr.1: 50535332-50535332 on Build GRCh38
rs763079271 Chr.1: 50535337-50535337 on Build GRCh38
rs763085352 Chr.1: 50535321-50535321 on Build GRCh38
rs952525910 Chr.1: 50535421-50535421 on Build GRCh38
rs748274334 Chr.1: 50535348-50535348 on Build GRCh38
rs147590569 Chr.1: 50535278-50535278 on Build GRCh38
rs960285730 Chr.1: 50535314-50535314 on Build GRCh38
rs563715182 Chr.1: 50535279-50535279 on Build GRCh38
rs894841256 Chr.1: 50535333-50535333 on Build GRCh38
rs993008307 Chr.1: 50535316-50535316 on Build GRCh38
rs1025446342 Chr.1: 50535415-50535415 on Build GRCh38
rs1053514551 Chr.1: 50535283-50535283 on Build GRCh38
rs994457294 Chr.1: 50535407-50535407 on Build GRCh38
rs911242364 Chr.1: 50535247-50535247 on Build GRCh38
rs757879012 Chr.1: 50535354-50535354 on Build GRCh38
rs531228972 Chr.1: 50535307-50535307 on Build GRCh38
rs946251200 Chr.1: 50535274-50535274 on Build GRCh38
rs922874865 Chr.1: 50535271-50535271 on Build GRCh38
rs754446068 Chr.1: 50535372-50535372 on Build GRCh38
rs936423586 Chr.1: 50535282-50535282 on Build GRCh38
rs751947091 Chr.1: 50535347-50535347 on Build GRCh38
rs370467557 Chr.1: 50535265-50535265 on Build GRCh38
rs373108605 Chr.1: 50535254-50535254 on Build GRCh38
rs202042404 Chr.1: 50535422-50535422 on Build GRCh38
rs764573723 Chr.1: 50535345-50535345 on Build GRCh38
rs374217344 Chr.1: 50535405-50535405 on Build GRCh38
rs376890368 Chr.1: 50535423-50535423 on Build GRCh38
rs773517723 Chr.1: 50535285-50535287 on Build GRCh38
rs578049886 Chr.1: 50535262-50535262 on Build GRCh38
rs753418578 Chr.1: 50535366-50535366 on Build GRCh38
rs778283291 Chr.1: 50535406-50535406 on Build GRCh38

View Full Product Details