Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95219862-95220131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM6
Entrez Gene ID
142827
Gene Name
acyl-CoA synthetase medium-chain family member 6
Gene Aliases
C10orf129, bA310E22.3
UniGene
Hs.134229
Gene Chromosome Location
Chr.10: 95194200-95228929 on Build GRCh38
Transcripts
NM_207321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1158791 c.1180T>C p.L394L Chr.10: 95219951-95219951 on Build GRCh38
921767 c.1131T>C p.C377C Chr.10: 95219902-95219902 on Build GRCh38
5371383 c.1128C>T p.L376L Chr.10: 95219899-95219899 on Build GRCh38
5371384 c.539C>T p.S180F Chr.10: 95219899-95219899 on Build GRCh38
3397326 c.1187C>T p.P396L Chr.10: 95219958-95219958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918532409 Chr.10: 95220038-95220038 on Build GRCh38
rs764533715 Chr.10: 95220068-95220068 on Build GRCh38
rs539543675 Chr.10: 95219892-95219892 on Build GRCh38
rs752861857 Chr.10: 95219933-95219933 on Build GRCh38
rs766519252 Chr.10: 95219906-95219906 on Build GRCh38
rs753683098 Chr.10: 95219951-95219951 on Build GRCh38
rs770277975 Chr.10: 95219997-95219997 on Build GRCh38
rs756097183 Chr.10: 95219943-95219943 on Build GRCh38
rs552821370 Chr.10: 95219913-95219913 on Build GRCh38
rs766388496 Chr.10: 95219946-95219948 on Build GRCh38
rs628102 Chr.10: 95220073-95220073 on Build GRCh38
rs773710442 Chr.10: 95220011-95220011 on Build GRCh38
rs757124613 Chr.10: 95219961-95219961 on Build GRCh38
rs11188229 Chr.10: 95220018-95220018 on Build GRCh38
rs763373086 Chr.10: 95219895-95219895 on Build GRCh38
rs771574975 Chr.10: 95219973-95219973 on Build GRCh38
rs763241107 Chr.10: 95220015-95220015 on Build GRCh38
rs767524744 Chr.10: 95219929-95219929 on Build GRCh38
rs746581821 Chr.10: 95219986-95219986 on Build GRCh38
rs769337864 Chr.10: 95219916-95219916 on Build GRCh38
rs762097283 Chr.10: 95220092-95220092 on Build GRCh38
rs774899760 Chr.10: 95219963-95219963 on Build GRCh38
rs1050814677 Chr.10: 95220089-95220089 on Build GRCh38
rs36042944 Chr.10: 95219978-95219979 on Build GRCh38
rs745532827 Chr.10: 95219964-95219964 on Build GRCh38
rs201299940 Chr.10: 95219944-95219944 on Build GRCh38
rs779403652 Chr.10: 95219983-95219983 on Build GRCh38

View Full Product Details