Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40860629-40860886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB7
Entrez Gene ID
150353
Gene Name
DnaJ heat shock protein family (Hsp40) member B7
Gene Aliases
DJ5, HSC3
UniGene
Hs.585042
Gene Chromosome Location
Chr.22: 40859549-40862126 on Build GRCh38
Transcripts
NM_145174.1 NM_001204827.1 NM_022098.3

Gene Symbol
XPNPEP3
Entrez Gene ID
63929
Gene Name
X-prolyl aminopeptidase 3
Gene Aliases
APP3, ICP55, NPHPL1
UniGene
Hs.529163
Gene Chromosome Location
Chr.22: 40857081-40932819 on Build GRCh38
Transcripts
NM_145174.1 NM_001204827.1 NM_022098.3

SNP Detail

SNP ID Chromosome Location
rs776646505 Chr.22: 40860825-40860826 on Build GRCh38
rs103197 Chr.22: 40860798-40860798 on Build GRCh38
rs115161413 Chr.22: 40860708-40860708 on Build GRCh38
rs975923013 Chr.22: 40860730-40860730 on Build GRCh38
rs550634591 Chr.22: 40860797-40860797 on Build GRCh38
rs554654980 Chr.22: 40860841-40860841 on Build GRCh38
rs748586679 Chr.22: 40860657-40860658 on Build GRCh38
rs531420664 Chr.22: 40860733-40860733 on Build GRCh38
rs1017944599 Chr.22: 40860718-40860718 on Build GRCh38
rs202011264 Chr.22: 40860657-40860657 on Build GRCh38
rs916849241 Chr.22: 40860838-40860838 on Build GRCh38
rs566902754 Chr.22: 40860842-40860842 on Build GRCh38
rs559236028 Chr.22: 40860845-40860845 on Build GRCh38
rs141950713 Chr.22: 40860721-40860721 on Build GRCh38
rs746110378 Chr.22: 40860811-40860811 on Build GRCh38
rs371177723 Chr.22: 40860737-40860737 on Build GRCh38
rs1006105238 Chr.22: 40860717-40860717 on Build GRCh38
rs976615474 Chr.22: 40860738-40860738 on Build GRCh38
rs888459959 Chr.22: 40860714-40860714 on Build GRCh38
rs966458046 Chr.22: 40860736-40860736 on Build GRCh38
rs767372693 Chr.22: 40860678-40860678 on Build GRCh38
rs749295521 Chr.22: 40860767-40860767 on Build GRCh38
rs1004936576 Chr.22: 40860688-40860688 on Build GRCh38
rs886453155 Chr.22: 40860662-40860662 on Build GRCh38
rs1026961875 Chr.22: 40860711-40860711 on Build GRCh38
rs747136589 Chr.22: 40860657-40860659 on Build GRCh38
rs756088018 Chr.22: 40860719-40860719 on Build GRCh38
rs777556440 Chr.22: 40860796-40860796 on Build GRCh38
rs536203870 Chr.22: 40860839-40860839 on Build GRCh38
rs1028231158 Chr.22: 40860746-40860746 on Build GRCh38
rs534204059 Chr.22: 40860843-40860843 on Build GRCh38
rs1030025027 Chr.22: 40860734-40860734 on Build GRCh38
rs936336491 Chr.22: 40860808-40860808 on Build GRCh38
rs5758111 Chr.22: 40860676-40860676 on Build GRCh38
rs897270723 Chr.22: 40860702-40860702 on Build GRCh38
rs751229782 Chr.22: 40860657-40860657 on Build GRCh38

View Full Product Details