Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45176316-45176541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADARB1
Entrez Gene ID
104
Gene Name
adenosine deaminase, RNA specific B1
Gene Aliases
ADAR2, DRABA2, DRADA2, RED1
UniGene
Hs.474018
Gene Chromosome Location
Chr.21: 45074578-45226563 on Build GRCh38
Transcripts
NM_001112.3 NM_001160230.1 NM_015833.3 NM_015834.3 NR_027672.1 NR_027673.1 NR_027674.1 NR_073200.1 XM_006723954.2 XM_011529425.2 XM_011529432.1 XM_017028242.1 XM_017028243.1 XM_017028244.1 XM_017028245.1 XM_017028246.1 XM_017028247.1 XM_017028248.1 XM_017028249.1 XM_017028250.1 XM_017028251.1 XM_017028252.1 XM_017028253.1 XM_017028254.1 XM_017028255.1 XM_017028256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4135048 c.720G>T p.L240F Chr.21: 45176421-45176421 on Build GRCh38
3551566 c.823G>A p.E275K Chr.21: 45176524-45176524 on Build GRCh38
1181978 c.731G>A p.R244H Chr.21: 45176432-45176432 on Build GRCh38
3551562 c.691C>T p.P231S Chr.21: 45176392-45176392 on Build GRCh38
6594678 c.730C>T p.R244C Chr.21: 45176431-45176431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748489139 Chr.21: 45176399-45176399 on Build GRCh38
rs766542655 Chr.21: 45176466-45176466 on Build GRCh38
rs755749136 Chr.21: 45176498-45176498 on Build GRCh38
rs977151734 Chr.21: 45176366-45176366 on Build GRCh38
rs200567642 Chr.21: 45176460-45176460 on Build GRCh38
rs750727660 Chr.21: 45176350-45176350 on Build GRCh38
rs73384785 Chr.21: 45176454-45176454 on Build GRCh38
rs139263374 Chr.21: 45176490-45176490 on Build GRCh38
rs139706517 Chr.21: 45176424-45176424 on Build GRCh38
rs753869362 Chr.21: 45176369-45176369 on Build GRCh38
rs200222908 Chr.21: 45176336-45176336 on Build GRCh38
rs765702168 Chr.21: 45176470-45176470 on Build GRCh38
rs756135135 Chr.21: 45176356-45176356 on Build GRCh38
rs751872032 Chr.21: 45176493-45176493 on Build GRCh38
rs774004725 Chr.21: 45176336-45176344 on Build GRCh38
rs868504673 Chr.21: 45176393-45176393 on Build GRCh38
rs745588497 Chr.21: 45176428-45176428 on Build GRCh38
rs530540302 Chr.21: 45176436-45176436 on Build GRCh38
rs769342205 Chr.21: 45176431-45176431 on Build GRCh38
rs772518113 Chr.21: 45176410-45176410 on Build GRCh38
rs534554822 Chr.21: 45176505-45176505 on Build GRCh38
rs778985197 Chr.21: 45176520-45176520 on Build GRCh38
rs370937062 Chr.21: 45176509-45176509 on Build GRCh38
rs779624932 Chr.21: 45176499-45176499 on Build GRCh38
rs766424463 Chr.21: 45176358-45176358 on Build GRCh38
rs942684350 Chr.21: 45176355-45176355 on Build GRCh38
rs150844459 Chr.21: 45176478-45176478 on Build GRCh38
rs539866699 Chr.21: 45176346-45176346 on Build GRCh38
rs758744746 Chr.21: 45176479-45176479 on Build GRCh38
rs774995629 Chr.21: 45176432-45176432 on Build GRCh38
rs753903571 Chr.21: 45176467-45176467 on Build GRCh38
rs925720923 Chr.21: 45176425-45176425 on Build GRCh38
rs772897635 Chr.21: 45176461-45176461 on Build GRCh38
rs755440291 Chr.21: 45176373-45176373 on Build GRCh38
rs145235270 Chr.21: 45176368-45176368 on Build GRCh38
rs922920611 Chr.21: 45176392-45176392 on Build GRCh38
rs932355965 Chr.21: 45176414-45176414 on Build GRCh38
rs760886166 Chr.21: 45176465-45176465 on Build GRCh38
rs377458378 Chr.21: 45176394-45176394 on Build GRCh38
rs199697177 Chr.21: 45176372-45176372 on Build GRCh38

View Full Product Details