Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37182512-37182765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF6
Entrez Gene ID
114904
Gene Name
C1q and tumor necrosis factor related protein 6
Gene Aliases
CTFP6, CTRP6, ZACRP6
UniGene
Hs.22011
Gene Chromosome Location
Chr.22: 37180166-37199400 on Build GRCh38
Transcripts
NM_031910.3 NM_182486.1 XM_011529857.1 XM_017028569.1 XM_017028570.1 XM_017028571.1 XM_017028572.1 XM_017028573.1 XM_017028574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6468148 c.124G>C p.E42Q Chr.22: 37182529-37182529 on Build GRCh38
259780 c.496G>A p.E166K Chr.22: 37182529-37182529 on Build GRCh38
5726419 c.97G>A p.G33S Chr.22: 37182556-37182556 on Build GRCh38
4753029 c.425G>A p.R142H Chr.22: 37182600-37182600 on Build GRCh38
4667247 c.70+1G>A p.? Chr.22: 37182735-37182735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781322736 Chr.22: 37182609-37182609 on Build GRCh38
rs761398579 Chr.22: 37182565-37182565 on Build GRCh38
rs757706065 Chr.22: 37182690-37182690 on Build GRCh38
rs775522637 Chr.22: 37182615-37182615 on Build GRCh38
rs17812699 Chr.22: 37182612-37182612 on Build GRCh38
rs749960126 Chr.22: 37182570-37182570 on Build GRCh38
rs769725904 Chr.22: 37182554-37182554 on Build GRCh38
rs200773711 Chr.22: 37182628-37182628 on Build GRCh38
rs760873324 Chr.22: 37182603-37182603 on Build GRCh38
rs780134428 Chr.22: 37182717-37182717 on Build GRCh38
rs199987518 Chr.22: 37182641-37182641 on Build GRCh38
rs986273635 Chr.22: 37182616-37182616 on Build GRCh38
rs187585450 Chr.22: 37182593-37182593 on Build GRCh38
rs749335466 Chr.22: 37182728-37182728 on Build GRCh38
rs753359141 Chr.22: 37182661-37182661 on Build GRCh38
rs768593656 Chr.22: 37182619-37182619 on Build GRCh38
rs200894137 Chr.22: 37182600-37182600 on Build GRCh38
rs753306549 Chr.22: 37182577-37182577 on Build GRCh38
rs368815003 Chr.22: 37182620-37182620 on Build GRCh38
rs775469822 Chr.22: 37182556-37182556 on Build GRCh38
rs763839877 Chr.22: 37182559-37182559 on Build GRCh38
rs764604009 Chr.22: 37182682-37182682 on Build GRCh38
rs774160358 Chr.22: 37182562-37182562 on Build GRCh38
rs746083093 Chr.22: 37182611-37182611 on Build GRCh38
rs767143723 Chr.22: 37182568-37182568 on Build GRCh38
rs756293028 Chr.22: 37182698-37182698 on Build GRCh38
rs229519 Chr.22: 37182539-37182539 on Build GRCh38
rs113314556 Chr.22: 37182617-37182617 on Build GRCh38
rs763392569 Chr.22: 37182667-37182667 on Build GRCh38
rs762931158 Chr.22: 37182557-37182557 on Build GRCh38
rs370547311 Chr.22: 37182553-37182553 on Build GRCh38
rs934551489 Chr.22: 37182724-37182724 on Build GRCh38
rs569663804 Chr.22: 37182696-37182696 on Build GRCh38
rs1015846593 Chr.22: 37182579-37182579 on Build GRCh38
rs751874002 Chr.22: 37182689-37182689 on Build GRCh38
rs763118868 Chr.22: 37182730-37182730 on Build GRCh38
rs760382972 Chr.22: 37182638-37182638 on Build GRCh38
rs781520974 Chr.22: 37182695-37182695 on Build GRCh38
rs755754179 Chr.22: 37182576-37182576 on Build GRCh38
rs766504497 Chr.22: 37182670-37182670 on Build GRCh38
rs528699550 Chr.22: 37182540-37182540 on Build GRCh38
rs968956911 Chr.22: 37182627-37182627 on Build GRCh38
rs757431560 Chr.22: 37182601-37182601 on Build GRCh38
rs758907614 Chr.22: 37182592-37182592 on Build GRCh38
rs773060013 Chr.22: 37182634-37182634 on Build GRCh38
rs373652334 Chr.22: 37182569-37182569 on Build GRCh38
rs1021357531 Chr.22: 37182736-37182736 on Build GRCh38
rs768485701 Chr.22: 37182737-37182737 on Build GRCh38
rs761604907 Chr.22: 37182626-37182626 on Build GRCh38

View Full Product Details