Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84415386-84415645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2C2
Entrez Gene ID
9914
Gene Name
ATPase secretory pathway Ca2+ transporting 2
Gene Aliases
SPCA2
UniGene
Hs.6168
Gene Chromosome Location
Chr.16: 84368523-84465777 on Build GRCh38
Transcripts
NM_001286527.2 NM_001291454.1 NM_014861.3 XM_011523486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7249389 c.624+9T>C p.? Chr.16: 84415600-84415600 on Build GRCh38
472188 c.550C>A p.R184R Chr.16: 84415517-84415517 on Build GRCh38
974391 c.578C>T p.S193F Chr.16: 84415545-84415545 on Build GRCh38
7225749 c.588C>T p.I196I Chr.16: 84415555-84415555 on Build GRCh38
974390 c.553G>T p.E185* Chr.16: 84415520-84415520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374182719 Chr.16: 84415502-84415502 on Build GRCh38
rs992705005 Chr.16: 84415620-84415620 on Build GRCh38
rs767446516 Chr.16: 84415474-84415474 on Build GRCh38
rs781099240 Chr.16: 84415445-84415445 on Build GRCh38
rs766942850 Chr.16: 84415586-84415586 on Build GRCh38
rs749498883 Chr.16: 84415515-84415515 on Build GRCh38
rs368339547 Chr.16: 84415541-84415541 on Build GRCh38
rs773744684 Chr.16: 84415591-84415591 on Build GRCh38
rs532747467 Chr.16: 84415524-84415524 on Build GRCh38
rs760170030 Chr.16: 84415476-84415476 on Build GRCh38
rs201484573 Chr.16: 84415580-84415580 on Build GRCh38
rs762151046 Chr.16: 84415578-84415578 on Build GRCh38
rs368761870 Chr.16: 84415562-84415562 on Build GRCh38
rs758999990 Chr.16: 84415573-84415573 on Build GRCh38
rs747455857 Chr.16: 84415432-84415432 on Build GRCh38
rs369793367 Chr.16: 84415468-84415468 on Build GRCh38
rs182026380 Chr.16: 84415615-84415615 on Build GRCh38
rs3743650 Chr.16: 84415426-84415426 on Build GRCh38
rs377747879 Chr.16: 84415471-84415471 on Build GRCh38
rs760240858 Chr.16: 84415535-84415535 on Build GRCh38
rs3743649 Chr.16: 84415418-84415418 on Build GRCh38
rs369498768 Chr.16: 84415617-84415617 on Build GRCh38
rs572809896 Chr.16: 84415422-84415422 on Build GRCh38
rs370237612 Chr.16: 84415475-84415475 on Build GRCh38
rs765695925 Chr.16: 84415461-84415462 on Build GRCh38
rs747640593 Chr.16: 84415455-84415455 on Build GRCh38
rs756718687 Chr.16: 84415583-84415583 on Build GRCh38
rs199516981 Chr.16: 84415604-84415604 on Build GRCh38
rs773085423 Chr.16: 84415563-84415563 on Build GRCh38
rs780481460 Chr.16: 84415514-84415514 on Build GRCh38
rs756309929 Chr.16: 84415552-84415552 on Build GRCh38
rs753978405 Chr.16: 84415587-84415587 on Build GRCh38
rs951359053 Chr.16: 84415428-84415428 on Build GRCh38
rs371217997 Chr.16: 84415591-84415591 on Build GRCh38
rs376277132 Chr.16: 84415616-84415616 on Build GRCh38
rs750928016 Chr.16: 84415551-84415551 on Build GRCh38
rs757585577 Chr.16: 84415547-84415547 on Build GRCh38
rs776152408 Chr.16: 84415569-84415569 on Build GRCh38
rs771913574 Chr.16: 84415529-84415529 on Build GRCh38
rs977205586 Chr.16: 84415424-84415424 on Build GRCh38
rs761905178 Chr.16: 84415462-84415462 on Build GRCh38
rs757738355 Chr.16: 84415507-84415507 on Build GRCh38
rs200595572 Chr.16: 84415568-84415568 on Build GRCh38
rs561594223 Chr.16: 84415430-84415430 on Build GRCh38
rs559173173 Chr.16: 84415420-84415420 on Build GRCh38
rs561980766 Chr.16: 84415518-84415518 on Build GRCh38
rs202097549 Chr.16: 84415470-84415470 on Build GRCh38
rs746770145 Chr.16: 84415602-84415602 on Build GRCh38
rs759470553 Chr.16: 84415623-84415623 on Build GRCh38
rs530606360 Chr.16: 84415494-84415494 on Build GRCh38
rs371738364 Chr.16: 84415606-84415606 on Build GRCh38
rs746929974 Chr.16: 84415565-84415565 on Build GRCh38
rs769928759 Chr.16: 84415510-84415510 on Build GRCh38
rs775142505 Chr.16: 84415577-84415577 on Build GRCh38
rs201180799 Chr.16: 84415454-84415454 on Build GRCh38
rs3826145 Chr.16: 84415600-84415600 on Build GRCh38
rs550371299 Chr.16: 84415506-84415506 on Build GRCh38
rs935938924 Chr.16: 84415603-84415603 on Build GRCh38
rs768610905 Chr.16: 84415517-84415517 on Build GRCh38
rs917031907 Chr.16: 84415489-84415489 on Build GRCh38
rs372651788 Chr.16: 84415488-84415488 on Build GRCh38
rs763467407 Chr.16: 84415621-84415622 on Build GRCh38
rs375031013 Chr.16: 84415540-84415540 on Build GRCh38
rs766019949 Chr.16: 84415479-84415479 on Build GRCh38
rs764812534 Chr.16: 84415574-84415574 on Build GRCh38
rs767915422 Chr.16: 84415549-84415549 on Build GRCh38
rs758180139 Chr.16: 84415597-84415597 on Build GRCh38
rs1057262659 Chr.16: 84415486-84415487 on Build GRCh38
rs781690343 Chr.16: 84415508-84415508 on Build GRCh38
rs1041439673 Chr.16: 84415505-84415505 on Build GRCh38
rs770164721 Chr.16: 84415449-84415449 on Build GRCh38
rs745387773 Chr.16: 84415607-84415607 on Build GRCh38
rs770471969 Chr.16: 84415567-84415567 on Build GRCh38
rs188980019 Chr.16: 84415596-84415596 on Build GRCh38
rs747896612 Chr.16: 84415557-84415557 on Build GRCh38
rs3743651 Chr.16: 84415555-84415555 on Build GRCh38
rs201237199 Chr.16: 84415556-84415556 on Build GRCh38
rs771088551 Chr.16: 84415439-84415439 on Build GRCh38
rs750932659 Chr.16: 84415581-84415581 on Build GRCh38
rs779067996 Chr.16: 84415594-84415594 on Build GRCh38
rs773130916 Chr.16: 84415532-84415532 on Build GRCh38
rs745923433 Chr.16: 84415448-84415448 on Build GRCh38

View Full Product Details