Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8863154-8863399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO1
Entrez Gene ID
2023
Gene Name
enolase 1
Gene Aliases
ENO1L1, HEL-S-17, MPB1, NNE, PPH
UniGene
Hs.517145
Gene Chromosome Location
Chr.1: 8861000-8879092 on Build GRCh38
Transcripts
NM_001201483.1 NM_001428.3 XM_006710433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620758 c.1136C>T p.T379I Chr.1: 8863275-8863275 on Build GRCh38
6574040 c.1135_1138delACCT p.T379fs*>55 Chr.1: 8863273-8863276 on Build GRCh38
912881 c.1086_1096del11 p.N363fs*11 Chr.1: 8863315-8863325 on Build GRCh38
6654077 c.1081C>T p.Q361* Chr.1: 8863330-8863330 on Build GRCh38
4522233 c.1152G>A p.L384L Chr.1: 8863259-8863259 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576937282 Chr.1: 8863313-8863313 on Build GRCh38
rs752714658 Chr.1: 8863193-8863193 on Build GRCh38
rs892214091 Chr.1: 8863349-8863349 on Build GRCh38
rs761571066 Chr.1: 8863346-8863346 on Build GRCh38
rs777807212 Chr.1: 8863212-8863212 on Build GRCh38
rs772881804 Chr.1: 8863367-8863367 on Build GRCh38
rs971855355 Chr.1: 8863256-8863256 on Build GRCh38
rs750328073 Chr.1: 8863275-8863275 on Build GRCh38
rs151047591 Chr.1: 8863323-8863323 on Build GRCh38
rs113129233 Chr.1: 8863234-8863234 on Build GRCh38
rs369228563 Chr.1: 8863199-8863199 on Build GRCh38
rs200008125 Chr.1: 8863341-8863341 on Build GRCh38
rs559265086 Chr.1: 8863356-8863357 on Build GRCh38
rs373861710 Chr.1: 8863292-8863292 on Build GRCh38
rs55906160 Chr.1: 8863200-8863200 on Build GRCh38
rs569618357 Chr.1: 8863296-8863296 on Build GRCh38
rs7523767 Chr.1: 8863222-8863222 on Build GRCh38
rs755798840 Chr.1: 8863188-8863188 on Build GRCh38
rs767088145 Chr.1: 8863355-8863355 on Build GRCh38
rs751103350 Chr.1: 8863258-8863258 on Build GRCh38
rs1054547298 Chr.1: 8863328-8863328 on Build GRCh38
rs761485123 Chr.1: 8863270-8863270 on Build GRCh38
rs774084402 Chr.1: 8863338-8863338 on Build GRCh38
rs767145197 Chr.1: 8863273-8863273 on Build GRCh38
rs747148122 Chr.1: 8863214-8863214 on Build GRCh38
rs879022873 Chr.1: 8863232-8863233 on Build GRCh38
rs750242166 Chr.1: 8863174-8863174 on Build GRCh38
rs143592223 Chr.1: 8863293-8863293 on Build GRCh38
rs775950694 Chr.1: 8863231-8863231 on Build GRCh38
rs893111143 Chr.1: 8863373-8863373 on Build GRCh38
rs765431276 Chr.1: 8863210-8863210 on Build GRCh38
rs879013965 Chr.1: 8863342-8863343 on Build GRCh38
rs878941002 Chr.1: 8863343-8863344 on Build GRCh38
rs756099793 Chr.1: 8863282-8863282 on Build GRCh38
rs200520582 Chr.1: 8863225-8863225 on Build GRCh38
rs962068072 Chr.1: 8863244-8863244 on Build GRCh38
rs755472016 Chr.1: 8863299-8863299 on Build GRCh38
rs11544489 Chr.1: 8863252-8863252 on Build GRCh38
rs762479923 Chr.1: 8863243-8863243 on Build GRCh38
rs763965291 Chr.1: 8863280-8863281 on Build GRCh38
rs920220427 Chr.1: 8863257-8863257 on Build GRCh38
rs770370916 Chr.1: 8863312-8863312 on Build GRCh38
rs746317051 Chr.1: 8863311-8863311 on Build GRCh38
rs777055807 Chr.1: 8863223-8863223 on Build GRCh38
rs749723672 Chr.1: 8863315-8863315 on Build GRCh38
rs867631690 Chr.1: 8863213-8863215 on Build GRCh38
rs751387298 Chr.1: 8863355-8863355 on Build GRCh38
rs879220242 Chr.1: 8863345-8863345 on Build GRCh38
rs758533996 Chr.1: 8863291-8863291 on Build GRCh38
rs3206672 Chr.1: 8863288-8863288 on Build GRCh38
rs760500130 Chr.1: 8863372-8863372 on Build GRCh38
rs763859148 Chr.1: 8863250-8863250 on Build GRCh38
rs150781341 Chr.1: 8863297-8863297 on Build GRCh38
rs762734833 Chr.1: 8863205-8863207 on Build GRCh38
rs766049412 Chr.1: 8863283-8863283 on Build GRCh38
rs770001763 Chr.1: 8863228-8863228 on Build GRCh38
rs753708347 Chr.1: 8863284-8863284 on Build GRCh38
rs368706320 Chr.1: 8863278-8863278 on Build GRCh38

View Full Product Details