Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128903296-128903569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf45
Entrez Gene ID
219833
Gene Name
chromosome 11 open reading frame 45
Gene Aliases
-
UniGene
Hs.689860
Gene Chromosome Location
Chr.11: 128899565-128909520 on Build GRCh38
Transcripts
NM_001256088.1 NM_145013.2 NR_045767.1 XM_011542654.2 XM_011542655.2 NM_000890.3

Gene Symbol
KCNJ5
Entrez Gene ID
3762
Gene Name
potassium voltage-gated channel subfamily J member 5
Gene Aliases
CIR, GIRK4, KATP1, KIR3.4, LQT13
UniGene
Hs.444595
Gene Chromosome Location
Chr.11: 128891418-128921163 on Build GRCh38
Transcripts
NM_001256088.1 NM_145013.2 NR_045767.1 XM_011542654.2 XM_011542655.2 NM_000890.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4145529 c.263A>C p.H88P Chr.11: 128903385-128903385 on Build GRCh38
3445196 c.219A>G p.V73V Chr.11: 128903429-128903429 on Build GRCh38
428697 c.167delA p.N56fs*17 Chr.11: 128903481-128903481 on Build GRCh38
1977546 c.250G>A p.A84T Chr.11: 128903398-128903398 on Build GRCh38
7228908 c.137-7A>G p.? Chr.11: 128903518-128903518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145654826 Chr.11: 128903321-128903322 on Build GRCh38
rs1052051811 Chr.11: 128903400-128903400 on Build GRCh38
rs776007111 Chr.11: 128903463-128903463 on Build GRCh38
rs758323029 Chr.11: 128903396-128903396 on Build GRCh38
rs749983858 Chr.11: 128903497-128903497 on Build GRCh38
rs551226762 Chr.11: 128903347-128903347 on Build GRCh38
rs780416853 Chr.11: 128903444-128903444 on Build GRCh38
rs534590917 Chr.11: 128903331-128903331 on Build GRCh38
rs758321524 Chr.11: 128903321-128903321 on Build GRCh38
rs772256910 Chr.11: 128903452-128903452 on Build GRCh38
rs1017694692 Chr.11: 128903441-128903441 on Build GRCh38
rs775805645 Chr.11: 128903363-128903363 on Build GRCh38
rs373333885 Chr.11: 128903505-128903505 on Build GRCh38
rs759028161 Chr.11: 128903369-128903370 on Build GRCh38
rs753510788 Chr.11: 128903379-128903379 on Build GRCh38
rs746034960 Chr.11: 128903545-128903545 on Build GRCh38
rs747088075 Chr.11: 128903537-128903537 on Build GRCh38
rs747731884 Chr.11: 128903498-128903498 on Build GRCh38
rs147863203 Chr.11: 128903384-128903384 on Build GRCh38
rs773602544 Chr.11: 128903344-128903344 on Build GRCh38
rs376118611 Chr.11: 128903351-128903351 on Build GRCh38
rs764927459 Chr.11: 128903496-128903496 on Build GRCh38
rs950763017 Chr.11: 128903506-128903506 on Build GRCh38
rs369632264 Chr.11: 128903345-128903345 on Build GRCh38
rs373451837 Chr.11: 128903428-128903428 on Build GRCh38
rs751600271 Chr.11: 128903516-128903516 on Build GRCh38
rs756876892 Chr.11: 128903382-128903382 on Build GRCh38
rs756287411 Chr.11: 128903434-128903434 on Build GRCh38
rs747230513 Chr.11: 128903482-128903482 on Build GRCh38
rs766043058 Chr.11: 128903508-128903508 on Build GRCh38
rs536583683 Chr.11: 128903467-128903467 on Build GRCh38
rs746259029 Chr.11: 128903450-128903450 on Build GRCh38
rs1026297854 Chr.11: 128903494-128903494 on Build GRCh38
rs745837429 Chr.11: 128903319-128903319 on Build GRCh38
rs370054655 Chr.11: 128903436-128903436 on Build GRCh38
rs754938819 Chr.11: 128903517-128903517 on Build GRCh38
rs768859509 Chr.11: 128903507-128903507 on Build GRCh38
rs199658982 Chr.11: 128903352-128903352 on Build GRCh38
rs80286035 Chr.11: 128903323-128903323 on Build GRCh38
rs748222755 Chr.11: 128903328-128903328 on Build GRCh38
rs373099600 Chr.11: 128903346-128903346 on Build GRCh38
rs753957937 Chr.11: 128903519-128903519 on Build GRCh38
rs150247728 Chr.11: 128903443-128903443 on Build GRCh38
rs768204066 Chr.11: 128903470-128903470 on Build GRCh38
rs760181371 Chr.11: 128903366-128903366 on Build GRCh38
rs746906239 Chr.11: 128903432-128903432 on Build GRCh38
rs932655032 Chr.11: 128903411-128903411 on Build GRCh38
rs779205400 Chr.11: 128903446-128903446 on Build GRCh38
rs141065881 Chr.11: 128903395-128903395 on Build GRCh38
rs4937387 Chr.11: 128903518-128903518 on Build GRCh38
rs761435667 Chr.11: 128903495-128903495 on Build GRCh38
rs750980891 Chr.11: 128903350-128903350 on Build GRCh38
rs749562350 Chr.11: 128903438-128903438 on Build GRCh38
rs763554422 Chr.11: 128903374-128903374 on Build GRCh38
rs771180439 Chr.11: 128903440-128903440 on Build GRCh38
rs779202570 Chr.11: 128903541-128903541 on Build GRCh38
rs781193701 Chr.11: 128903324-128903324 on Build GRCh38
rs934932915 Chr.11: 128903369-128903369 on Build GRCh38

View Full Product Details