Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123785687-123785960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
UniGene
Hs.622654
Gene Chromosome Location
Chr.12: 123761995-123935720 on Build GRCh38
Transcripts
NM_207437.3 XM_005253555.3 XM_011538014.2 XM_011538015.2 XM_011538016.2 XM_011538017.2 XM_011538020.2 XM_017018960.1 XM_017018961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4442708 c.1127T>C p.I376T Chr.12: 123785825-123785825 on Build GRCh38
467958 c.577T>A p.W193R Chr.12: 123785821-123785821 on Build GRCh38
3457572 c.612G>A p.R204R Chr.12: 123785856-123785856 on Build GRCh38
5886775 c.1238+5G>A p.? Chr.12: 123785941-123785941 on Build GRCh38
5510649 c.607G>A p.E203K Chr.12: 123785851-123785851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894747559 Chr.12: 123785910-123785910 on Build GRCh38
rs139439839 Chr.12: 123785850-123785850 on Build GRCh38
rs769259387 Chr.12: 123785834-123785834 on Build GRCh38
rs148669304 Chr.12: 123785892-123785892 on Build GRCh38
rs778379989 Chr.12: 123785908-123785908 on Build GRCh38
rs199697349 Chr.12: 123785821-123785821 on Build GRCh38
rs763876955 Chr.12: 123785798-123785798 on Build GRCh38
rs370483689 Chr.12: 123785754-123785754 on Build GRCh38
rs770966397 Chr.12: 123785896-123785897 on Build GRCh38
rs775097365 Chr.12: 123785792-123785792 on Build GRCh38
rs769355576 Chr.12: 123785919-123785919 on Build GRCh38
rs774201564 Chr.12: 123785865-123785865 on Build GRCh38
rs752517159 Chr.12: 123785717-123785717 on Build GRCh38
rs767513669 Chr.12: 123785880-123785880 on Build GRCh38
rs568999192 Chr.12: 123785767-123785767 on Build GRCh38
rs748393634 Chr.12: 123785825-123785825 on Build GRCh38
rs779431805 Chr.12: 123785820-123785820 on Build GRCh38
rs957321467 Chr.12: 123785929-123785929 on Build GRCh38
rs116082071 Chr.12: 123785818-123785818 on Build GRCh38
rs1013484989 Chr.12: 123785916-123785916 on Build GRCh38
rs533763833 Chr.12: 123785807-123785807 on Build GRCh38
rs1023783431 Chr.12: 123785769-123785769 on Build GRCh38
rs74943885 Chr.12: 123785800-123785800 on Build GRCh38
rs764887430 Chr.12: 123785788-123785788 on Build GRCh38
rs767291816 Chr.12: 123785808-123785808 on Build GRCh38
rs532466984 Chr.12: 123785733-123785733 on Build GRCh38
rs749191930 Chr.12: 123785836-123785837 on Build GRCh38
rs756098229 Chr.12: 123785812-123785812 on Build GRCh38
rs777462563 Chr.12: 123785790-123785790 on Build GRCh38
rs79726540 Chr.12: 123785924-123785924 on Build GRCh38
rs374153461 Chr.12: 123785772-123785772 on Build GRCh38
rs370924488 Chr.12: 123785923-123785923 on Build GRCh38
rs761476141 Chr.12: 123785874-123785874 on Build GRCh38
rs747469941 Chr.12: 123785914-123785914 on Build GRCh38
rs182095656 Chr.12: 123785732-123785732 on Build GRCh38
rs368461067 Chr.12: 123785859-123785859 on Build GRCh38
rs763680952 Chr.12: 123785864-123785864 on Build GRCh38
rs373800707 Chr.12: 123785721-123785721 on Build GRCh38
rs762584820 Chr.12: 123785851-123785851 on Build GRCh38
rs979868924 Chr.12: 123785793-123785793 on Build GRCh38
rs761717192 Chr.12: 123785927-123785927 on Build GRCh38
rs878907296 Chr.12: 123785869-123785869 on Build GRCh38
rs758927807 Chr.12: 123785785-123785785 on Build GRCh38
rs753125598 Chr.12: 123785898-123785898 on Build GRCh38
rs762780876 Chr.12: 123785794-123785794 on Build GRCh38
rs200542157 Chr.12: 123785833-123785833 on Build GRCh38
rs202034758 Chr.12: 123785722-123785722 on Build GRCh38
rs560209246 Chr.12: 123785930-123785930 on Build GRCh38
rs144167895 Chr.12: 123785753-123785753 on Build GRCh38
rs147159744 Chr.12: 123785889-123785889 on Build GRCh38
rs143556029 Chr.12: 123785881-123785881 on Build GRCh38
rs750550022 Chr.12: 123785809-123785809 on Build GRCh38
rs138303428 Chr.12: 123785763-123785763 on Build GRCh38
rs566620103 Chr.12: 123785806-123785806 on Build GRCh38
rs764727037 Chr.12: 123785715-123785715 on Build GRCh38
rs757350192 Chr.12: 123785728-123785728 on Build GRCh38
rs769109966 Chr.12: 123785757-123785757 on Build GRCh38
rs758664225 Chr.12: 123785906-123785906 on Build GRCh38
rs778959839 Chr.12: 123785893-123785893 on Build GRCh38
rs76470273 Chr.12: 123785877-123785877 on Build GRCh38
rs775064137 Chr.12: 123785758-123785758 on Build GRCh38
rs995378362 Chr.12: 123785719-123785719 on Build GRCh38
rs749515621 Chr.12: 123785813-123785813 on Build GRCh38
rs772661844 Chr.12: 123785830-123785830 on Build GRCh38
rs766553703 Chr.12: 123785884-123785884 on Build GRCh38
rs116310078 Chr.12: 123785920-123785920 on Build GRCh38
rs78912656 Chr.12: 123785879-123785879 on Build GRCh38
rs944806735 Chr.12: 123785875-123785875 on Build GRCh38
rs147774367 Chr.12: 123785801-123785801 on Build GRCh38
rs945956782 Chr.12: 123785816-123785816 on Build GRCh38
rs769568814 Chr.12: 123785713-123785714 on Build GRCh38
rs78625381 Chr.12: 123785789-123785789 on Build GRCh38

View Full Product Details