Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50751441-50751702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLD
Entrez Gene ID
1540
Gene Name
CYLD lysine 63 deubiquitinase
Gene Aliases
BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2
UniGene
Hs.578973
Gene Chromosome Location
Chr.16: 50742026-50801935 on Build GRCh38
Transcripts
NM_001042355.1 NM_001042412.1 NM_015247.2 XM_005255812.2 XM_006721149.1 XM_011522907.2 XM_011522908.1 XM_011522911.2 XM_017022977.1 XM_017022978.1 XM_017022979.1 XM_017022980.1 XM_017022981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363376 c.557T>C p.L186P Chr.16: 50751656-50751656 on Build GRCh38
5363375 c.557T>C p.L186P Chr.16: 50751656-50751656 on Build GRCh38
5646273 c.528C>T p.F176F Chr.16: 50751627-50751627 on Build GRCh38
6638976 c.515G>A p.R172H Chr.16: 50751614-50751614 on Build GRCh38
3957632 c.540G>T p.V180V Chr.16: 50751639-50751639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899723683 Chr.16: 50751551-50751551 on Build GRCh38
rs192488410 Chr.16: 50751561-50751561 on Build GRCh38
rs370450747 Chr.16: 50751637-50751637 on Build GRCh38
rs1053844205 Chr.16: 50751488-50751488 on Build GRCh38
rs374118770 Chr.16: 50751668-50751668 on Build GRCh38
rs781763284 Chr.16: 50751574-50751577 on Build GRCh38
rs763461167 Chr.16: 50751584-50751584 on Build GRCh38
rs869245135 Chr.16: 50751599-50751599 on Build GRCh38
rs746877025 Chr.16: 50751562-50751562 on Build GRCh38
rs757203445 Chr.16: 50751663-50751663 on Build GRCh38
rs201836260 Chr.16: 50751613-50751613 on Build GRCh38
rs777360812 Chr.16: 50751653-50751653 on Build GRCh38
rs758164507 Chr.16: 50751560-50751560 on Build GRCh38
rs12599808 Chr.16: 50751617-50751617 on Build GRCh38
rs200230348 Chr.16: 50751575-50751575 on Build GRCh38
rs371024626 Chr.16: 50751589-50751589 on Build GRCh38
rs752294416 Chr.16: 50751642-50751642 on Build GRCh38
rs71384575 Chr.16: 50751477-50751477 on Build GRCh38
rs377383082 Chr.16: 50751633-50751633 on Build GRCh38
rs965699210 Chr.16: 50751616-50751616 on Build GRCh38
rs753356127 Chr.16: 50751634-50751634 on Build GRCh38
rs201431389 Chr.16: 50751681-50751681 on Build GRCh38
rs758217895 Chr.16: 50751649-50751649 on Build GRCh38
rs953229985 Chr.16: 50751586-50751586 on Build GRCh38
rs1040027011 Chr.16: 50751545-50751545 on Build GRCh38
rs892283137 Chr.16: 50751573-50751573 on Build GRCh38
rs780671666 Chr.16: 50751554-50751554 on Build GRCh38
rs1012947567 Chr.16: 50751543-50751543 on Build GRCh38
rs986490098 Chr.16: 50751606-50751606 on Build GRCh38
rs747564453 Chr.16: 50751557-50751557 on Build GRCh38
rs764917904 Chr.16: 50751639-50751639 on Build GRCh38
rs771043455 Chr.16: 50751563-50751563 on Build GRCh38
rs781078779 Chr.16: 50751569-50751569 on Build GRCh38
rs894066967 Chr.16: 50751539-50751539 on Build GRCh38
rs751525184 Chr.16: 50751654-50751654 on Build GRCh38
rs888478270 Chr.16: 50751540-50751540 on Build GRCh38
rs745934140 Chr.16: 50751572-50751572 on Build GRCh38

View Full Product Details