Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50193098-50193359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT1
Entrez Gene ID
2779
Gene Name
G protein subunit alpha transducin 1
Gene Aliases
CSNB1G, CSNBAD3, GBT1, GNATR
UniGene
Hs.517978
Gene Chromosome Location
Chr.3: 50191610-50197696 on Build GRCh38
Transcripts
NM_000172.3 NM_144499.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116906 c.139A>C p.K47Q Chr.3: 50193165-50193165 on Build GRCh38
6165078 c.129C>T p.S43S Chr.3: 50193155-50193155 on Build GRCh38
4953340 c.213C>A p.G71G Chr.3: 50193328-50193328 on Build GRCh38
7369647 c.141G>T p.K47N Chr.3: 50193167-50193167 on Build GRCh38
269252 c.224A>G p.Q75R Chr.3: 50193339-50193339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs104893740 Chr.3: 50193139-50193139 on Build GRCh38
rs747846195 Chr.3: 50193283-50193283 on Build GRCh38
rs776367777 Chr.3: 50193152-50193152 on Build GRCh38
rs376261970 Chr.3: 50193238-50193238 on Build GRCh38
rs34797487 Chr.3: 50193143-50193143 on Build GRCh38
rs772024349 Chr.3: 50193133-50193133 on Build GRCh38
rs200425911 Chr.3: 50193201-50193201 on Build GRCh38
rs749921670 Chr.3: 50193168-50193168 on Build GRCh38
rs201449427 Chr.3: 50193248-50193248 on Build GRCh38
rs758300344 Chr.3: 50193234-50193234 on Build GRCh38
rs778486199 Chr.3: 50193275-50193275 on Build GRCh38
rs762489106 Chr.3: 50193325-50193325 on Build GRCh38
rs745833016 Chr.3: 50193122-50193122 on Build GRCh38
rs768025180 Chr.3: 50193191-50193191 on Build GRCh38
rs930177439 Chr.3: 50193256-50193256 on Build GRCh38
rs745911529 Chr.3: 50193233-50193233 on Build GRCh38
rs779976608 Chr.3: 50193235-50193235 on Build GRCh38
rs750374101 Chr.3: 50193254-50193254 on Build GRCh38
rs754812661 Chr.3: 50193209-50193209 on Build GRCh38
rs190126440 Chr.3: 50193286-50193286 on Build GRCh38
rs990570090 Chr.3: 50193115-50193115 on Build GRCh38
rs760015293 Chr.3: 50193187-50193187 on Build GRCh38
rs772624047 Chr.3: 50193289-50193289 on Build GRCh38
rs781505899 Chr.3: 50193211-50193212 on Build GRCh38
rs951812811 Chr.3: 50193176-50193176 on Build GRCh38
rs770010487 Chr.3: 50193219-50193220 on Build GRCh38
rs776116147 Chr.3: 50193329-50193329 on Build GRCh38
rs764910362 Chr.3: 50193162-50193162 on Build GRCh38
rs745721123 Chr.3: 50193194-50193196 on Build GRCh38
rs1041904269 Chr.3: 50193278-50193278 on Build GRCh38
rs562770722 Chr.3: 50193201-50193201 on Build GRCh38
rs767935708 Chr.3: 50193326-50193326 on Build GRCh38
rs778648409 Chr.3: 50193119-50193123 on Build GRCh38
rs776566245 Chr.3: 50193280-50193280 on Build GRCh38
rs761142046 Chr.3: 50193340-50193340 on Build GRCh38
rs768429545 Chr.3: 50193147-50193147 on Build GRCh38
rs778094885 Chr.3: 50193222-50193222 on Build GRCh38
rs371336879 Chr.3: 50193192-50193192 on Build GRCh38
rs371980223 Chr.3: 50193223-50193223 on Build GRCh38
rs374528386 Chr.3: 50193204-50193204 on Build GRCh38
rs757438553 Chr.3: 50193224-50193224 on Build GRCh38
rs778959098 Chr.3: 50193119-50193119 on Build GRCh38
rs753835927 Chr.3: 50193126-50193126 on Build GRCh38
rs778908618 Chr.3: 50193230-50193230 on Build GRCh38

View Full Product Details