Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50320291-50320542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL2
Entrez Gene ID
8692
Gene Name
hyaluronoglucosaminidase 2
Gene Aliases
LUCA2
UniGene
Hs.76873
Gene Chromosome Location
Chr.3: 50317790-50322850 on Build GRCh38
Transcripts
NM_003773.4 NM_033158.4 XM_005265524.2 XM_005265525.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1162360 c.52T>G p.S18A Chr.3: 50320438-50320438 on Build GRCh38
6296208 c.90C>A p.I30I Chr.3: 50320400-50320400 on Build GRCh38
1046483 c.46_47insG p.A16fs*46 Chr.3: 50320443-50320444 on Build GRCh38
4152471 c.192C>G p.A64A Chr.3: 50320298-50320298 on Build GRCh38
6221076 c.177C>T p.A59A Chr.3: 50320313-50320313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048448022 Chr.3: 50320462-50320462 on Build GRCh38
rs7629425 Chr.3: 50320498-50320498 on Build GRCh38
rs587755085 Chr.3: 50320334-50320334 on Build GRCh38
rs782305305 Chr.3: 50320445-50320445 on Build GRCh38
rs781979383 Chr.3: 50320344-50320344 on Build GRCh38
rs782321507 Chr.3: 50320324-50320324 on Build GRCh38
rs709210 Chr.3: 50320438-50320438 on Build GRCh38
rs781960766 Chr.3: 50320313-50320313 on Build GRCh38
rs781826287 Chr.3: 50320406-50320406 on Build GRCh38
rs200709569 Chr.3: 50320331-50320331 on Build GRCh38
rs755417 Chr.3: 50320504-50320504 on Build GRCh38
rs782207038 Chr.3: 50320321-50320321 on Build GRCh38
rs782135520 Chr.3: 50320466-50320466 on Build GRCh38
rs782089963 Chr.3: 50320483-50320483 on Build GRCh38
rs782667176 Chr.3: 50320509-50320509 on Build GRCh38
rs782531574 Chr.3: 50320411-50320412 on Build GRCh38
rs782629042 Chr.3: 50320403-50320403 on Build GRCh38
rs782035304 Chr.3: 50320451-50320451 on Build GRCh38
rs587651461 Chr.3: 50320424-50320424 on Build GRCh38
rs897777709 Chr.3: 50320506-50320506 on Build GRCh38
rs781802881 Chr.3: 50320379-50320379 on Build GRCh38
rs782567680 Chr.3: 50320412-50320412 on Build GRCh38
rs782471279 Chr.3: 50320407-50320407 on Build GRCh38
rs373163483 Chr.3: 50320322-50320322 on Build GRCh38
rs782060503 Chr.3: 50320443-50320443 on Build GRCh38
rs781791940 Chr.3: 50320521-50320521 on Build GRCh38
rs782125730 Chr.3: 50320309-50320309 on Build GRCh38
rs1004190508 Chr.3: 50320492-50320492 on Build GRCh38
rs113495715 Chr.3: 50320490-50320490 on Build GRCh38
rs782772862 Chr.3: 50320341-50320341 on Build GRCh38
rs368920956 Chr.3: 50320513-50320513 on Build GRCh38
rs782402304 Chr.3: 50320429-50320429 on Build GRCh38
rs782712109 Chr.3: 50320485-50320485 on Build GRCh38
rs782095564 Chr.3: 50320370-50320370 on Build GRCh38
rs782765717 Chr.3: 50320469-50320469 on Build GRCh38
rs782763712 Chr.3: 50320400-50320400 on Build GRCh38
rs371492352 Chr.3: 50320402-50320402 on Build GRCh38
rs376665287 Chr.3: 50320385-50320385 on Build GRCh38
rs781855735 Chr.3: 50320495-50320496 on Build GRCh38
rs782819086 Chr.3: 50320493-50320493 on Build GRCh38
rs996909074 Chr.3: 50320461-50320461 on Build GRCh38
rs782644575 Chr.3: 50320431-50320431 on Build GRCh38
rs781845744 Chr.3: 50320473-50320473 on Build GRCh38
rs1050072 Chr.3: 50320499-50320499 on Build GRCh38
rs782500260 Chr.3: 50320514-50320514 on Build GRCh38
rs782718260 Chr.3: 50320375-50320375 on Build GRCh38
rs781951272 Chr.3: 50320442-50320442 on Build GRCh38

View Full Product Details