Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27445141-27445384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT172
Entrez Gene ID
26160
Gene Name
intraflagellar transport 172
Gene Aliases
BBS20, NPHP17, RP71, SLB, SRTD10, osm-1, wim
UniGene
Hs.127401
Gene Chromosome Location
Chr.2: 27444373-27489819 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
UniGene
Hs.59509
Gene Chromosome Location
Chr.2: 27442366-27446481 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1407461 c.5050C>A p.L1684M Chr.2: 27445314-27445314 on Build GRCh38
1191853 c.4994A>G p.D1665G Chr.2: 27445370-27445370 on Build GRCh38
281863 c.5044C>T p.R1682* Chr.2: 27445320-27445320 on Build GRCh38
6395345 c.5013G>T p.E1671D Chr.2: 27445351-27445351 on Build GRCh38
1172865 c.5027C>T p.A1676V Chr.2: 27445337-27445337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755880203 Chr.2: 27445255-27445255 on Build GRCh38
rs761028024 Chr.2: 27445360-27445360 on Build GRCh38
rs748486924 Chr.2: 27445312-27445312 on Build GRCh38
rs1033492381 Chr.2: 27445240-27445240 on Build GRCh38
rs377082478 Chr.2: 27445272-27445272 on Build GRCh38
rs564816832 Chr.2: 27445183-27445183 on Build GRCh38
rs201017771 Chr.2: 27445364-27445364 on Build GRCh38
rs774361785 Chr.2: 27445340-27445340 on Build GRCh38
rs916432121 Chr.2: 27445178-27445178 on Build GRCh38
rs200049734 Chr.2: 27445296-27445296 on Build GRCh38
rs931222015 Chr.2: 27445176-27445176 on Build GRCh38
rs778545853 Chr.2: 27445275-27445275 on Build GRCh38
rs752473707 Chr.2: 27445246-27445246 on Build GRCh38
rs972056548 Chr.2: 27445331-27445331 on Build GRCh38
rs759639938 Chr.2: 27445342-27445342 on Build GRCh38
rs949178000 Chr.2: 27445235-27445235 on Build GRCh38
rs878962267 Chr.2: 27445168-27445168 on Build GRCh38
rs919877709 Chr.2: 27445176-27445176 on Build GRCh38
rs375383466 Chr.2: 27445269-27445269 on Build GRCh38
rs774447797 Chr.2: 27445188-27445188 on Build GRCh38
rs773691500 Chr.2: 27445319-27445319 on Build GRCh38
rs532307393 Chr.2: 27445232-27445232 on Build GRCh38
rs749180868 Chr.2: 27445270-27445270 on Build GRCh38
rs543062555 Chr.2: 27445177-27445177 on Build GRCh38
rs776071272 Chr.2: 27445283-27445283 on Build GRCh38
rs745393421 Chr.2: 27445280-27445280 on Build GRCh38
rs979602305 Chr.2: 27445215-27445215 on Build GRCh38
rs917873749 Chr.2: 27445305-27445305 on Build GRCh38
rs575981350 Chr.2: 27445174-27445174 on Build GRCh38
rs763335302 Chr.2: 27445323-27445323 on Build GRCh38
rs770040628 Chr.2: 27445313-27445313 on Build GRCh38
rs757212554 Chr.2: 27445365-27445365 on Build GRCh38
rs199786896 Chr.2: 27445260-27445260 on Build GRCh38
rs775285304 Chr.2: 27445311-27445311 on Build GRCh38
rs777683967 Chr.2: 27445259-27445259 on Build GRCh38
rs1046597191 Chr.2: 27445245-27445245 on Build GRCh38
rs766918099 Chr.2: 27445335-27445335 on Build GRCh38
rs767672341 Chr.2: 27445346-27445346 on Build GRCh38
rs146575848 Chr.2: 27445359-27445359 on Build GRCh38
rs959229695 Chr.2: 27445273-27445273 on Build GRCh38

View Full Product Details