Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80884132-80884405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEMIP
Entrez Gene ID
57214
Gene Name
cell migration inducing hyaluronan binding protein
Gene Aliases
CCSP1, HYBID, KIAA1199, TMEM2L
UniGene
Hs.459088
Gene Chromosome Location
Chr.15: 80779343-80951662 on Build GRCh38
Transcripts
NM_001293298.1 NM_001293304.1 NM_018689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
434399 c.630T>C p.Y210Y Chr.15: 80884187-80884187 on Build GRCh38
161935 c.715G>T p.E239* Chr.15: 80884272-80884272 on Build GRCh38
4505637 c.698C>T p.S233F Chr.15: 80884255-80884255 on Build GRCh38
3504461 c.693C>T p.I231I Chr.15: 80884250-80884250 on Build GRCh38
3504462 c.786C>T p.H262H Chr.15: 80884343-80884343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562341405 Chr.15: 80884313-80884313 on Build GRCh38
rs181470477 Chr.15: 80884239-80884239 on Build GRCh38
rs756702391 Chr.15: 80884276-80884277 on Build GRCh38
rs74625258 Chr.15: 80884357-80884357 on Build GRCh38
rs112282520 Chr.15: 80884285-80884285 on Build GRCh38
rs779454573 Chr.15: 80884240-80884240 on Build GRCh38
rs760669306 Chr.15: 80884276-80884276 on Build GRCh38
rs143215956 Chr.15: 80884189-80884189 on Build GRCh38
rs763699954 Chr.15: 80884156-80884157 on Build GRCh38
rs781169090 Chr.15: 80884254-80884254 on Build GRCh38
rs745937898 Chr.15: 80884295-80884295 on Build GRCh38
rs771084010 Chr.15: 80884281-80884281 on Build GRCh38
rs34765306 Chr.15: 80884172-80884172 on Build GRCh38
rs778668213 Chr.15: 80884277-80884278 on Build GRCh38
rs1033739108 Chr.15: 80884344-80884344 on Build GRCh38
rs763926290 Chr.15: 80884219-80884219 on Build GRCh38
rs906522260 Chr.15: 80884306-80884306 on Build GRCh38
rs973250044 Chr.15: 80884180-80884180 on Build GRCh38
rs943062300 Chr.15: 80884225-80884225 on Build GRCh38
rs375708015 Chr.15: 80884238-80884238 on Build GRCh38
rs151251518 Chr.15: 80884202-80884202 on Build GRCh38
rs202247286 Chr.15: 80884200-80884200 on Build GRCh38
rs754128576 Chr.15: 80884197-80884197 on Build GRCh38
rs750412324 Chr.15: 80884320-80884320 on Build GRCh38
rs766497726 Chr.15: 80884190-80884190 on Build GRCh38
rs201227565 Chr.15: 80884265-80884265 on Build GRCh38
rs202049796 Chr.15: 80884231-80884231 on Build GRCh38
rs911556696 Chr.15: 80884210-80884210 on Build GRCh38
rs759784850 Chr.15: 80884297-80884297 on Build GRCh38
rs751367288 Chr.15: 80884309-80884309 on Build GRCh38
rs35343835 Chr.15: 80884380-80884380 on Build GRCh38
rs761725082 Chr.15: 80884310-80884310 on Build GRCh38
rs1048497673 Chr.15: 80884206-80884206 on Build GRCh38
rs201209391 Chr.15: 80884371-80884371 on Build GRCh38
rs754686368 Chr.15: 80884373-80884373 on Build GRCh38
rs111716189 Chr.15: 80884230-80884230 on Build GRCh38
rs773173007 Chr.15: 80884162-80884162 on Build GRCh38
rs140482625 Chr.15: 80884229-80884229 on Build GRCh38
rs753346475 Chr.15: 80884274-80884275 on Build GRCh38
rs767597905 Chr.15: 80884157-80884157 on Build GRCh38
rs542993564 Chr.15: 80884303-80884303 on Build GRCh38
rs755553701 Chr.15: 80884353-80884353 on Build GRCh38
rs753504492 Chr.15: 80884372-80884372 on Build GRCh38
rs772472795 Chr.15: 80884261-80884261 on Build GRCh38
rs776812160 Chr.15: 80884282-80884282 on Build GRCh38
rs755714140 Chr.15: 80884232-80884232 on Build GRCh38
rs1053892693 Chr.15: 80884246-80884246 on Build GRCh38
rs748602893 Chr.15: 80884250-80884250 on Build GRCh38

View Full Product Details