Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94073200-94073465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPHL1
Entrez Gene ID
341208
Gene Name
hephaestin like 1
Gene Aliases
-
UniGene
Hs.669645
Gene Chromosome Location
Chr.11: 94021212-94114208 on Build GRCh38
Transcripts
NM_001098672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913202 c.1504+1G>A p.? Chr.11: 94073440-94073440 on Build GRCh38
5739417 c.1421C>T p.A474V Chr.11: 94073356-94073356 on Build GRCh38
933451 c.1455C>T p.P485P Chr.11: 94073390-94073390 on Build GRCh38
4690011 c.1399delG p.D467fs*4 Chr.11: 94073334-94073334 on Build GRCh38
933450 c.1445G>T p.S482I Chr.11: 94073380-94073380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767910593 Chr.11: 94073369-94073369 on Build GRCh38
rs377529278 Chr.11: 94073306-94073306 on Build GRCh38
rs1045614251 Chr.11: 94073408-94073408 on Build GRCh38
rs570447213 Chr.11: 94073368-94073368 on Build GRCh38
rs963976717 Chr.11: 94073422-94073422 on Build GRCh38
rs754920169 Chr.11: 94073399-94073399 on Build GRCh38
rs937570756 Chr.11: 94073235-94073235 on Build GRCh38
rs772844212 Chr.11: 94073331-94073331 on Build GRCh38
rs1007906109 Chr.11: 94073402-94073402 on Build GRCh38
rs886819086 Chr.11: 94073397-94073397 on Build GRCh38
rs371055898 Chr.11: 94073441-94073441 on Build GRCh38
rs777484309 Chr.11: 94073412-94073412 on Build GRCh38
rs763396661 Chr.11: 94073342-94073342 on Build GRCh38
rs747999009 Chr.11: 94073405-94073405 on Build GRCh38
rs746160001 Chr.11: 94073285-94073287 on Build GRCh38
rs995699162 Chr.11: 94073328-94073328 on Build GRCh38
rs377516597 Chr.11: 94073296-94073296 on Build GRCh38
rs764643299 Chr.11: 94073353-94073353 on Build GRCh38
rs768597758 Chr.11: 94073298-94073298 on Build GRCh38
rs549708756 Chr.11: 94073340-94073340 on Build GRCh38
rs780125027 Chr.11: 94073284-94073284 on Build GRCh38
rs781534264 Chr.11: 94073261-94073261 on Build GRCh38
rs1914725 Chr.11: 94073250-94073250 on Build GRCh38
rs759027579 Chr.11: 94073426-94073426 on Build GRCh38
rs766398855 Chr.11: 94073396-94073396 on Build GRCh38
rs752011824 Chr.11: 94073366-94073366 on Build GRCh38
rs760232781 Chr.11: 94073336-94073336 on Build GRCh38
rs907028130 Chr.11: 94073419-94073419 on Build GRCh38
rs778787587 Chr.11: 94073400-94073400 on Build GRCh38
rs779110608 Chr.11: 94073259-94073260 on Build GRCh38
rs753905528 Chr.11: 94073398-94073398 on Build GRCh38
rs867677199 Chr.11: 94073394-94073394 on Build GRCh38
rs568612189 Chr.11: 94073433-94073433 on Build GRCh38
rs371001785 Chr.11: 94073316-94073316 on Build GRCh38
rs756291743 Chr.11: 94073379-94073379 on Build GRCh38
rs772508437 Chr.11: 94073311-94073312 on Build GRCh38
rs778701610 Chr.11: 94073299-94073299 on Build GRCh38
rs771747675 Chr.11: 94073327-94073327 on Build GRCh38
rs375747139 Chr.11: 94073376-94073376 on Build GRCh38
rs985608725 Chr.11: 94073240-94073240 on Build GRCh38
rs928903587 Chr.11: 94073233-94073233 on Build GRCh38
rs776169161 Chr.11: 94073341-94073341 on Build GRCh38
rs200956976 Chr.11: 94073290-94073290 on Build GRCh38
rs955192888 Chr.11: 94073229-94073229 on Build GRCh38
rs750597871 Chr.11: 94073266-94073266 on Build GRCh38
rs372472920 Chr.11: 94073273-94073273 on Build GRCh38
rs750689503 Chr.11: 94073373-94073373 on Build GRCh38

View Full Product Details