Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49747861-49748098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K1
Entrez Gene ID
9807
Gene Name
inositol hexakisphosphate kinase 1
Gene Aliases
IHPK1, PiUS
UniGene
Hs.386168
Gene Chromosome Location
Chr.3: 49724295-49786540 on Build GRCh38
Transcripts
NM_001006115.2 NM_001242829.1 NM_153273.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4774552 c.161G>A p.R54Q Chr.3: 49747880-49747880 on Build GRCh38
6404026 c.11G>T p.C4F Chr.3: 49748030-49748030 on Build GRCh38
1291729 c.118A>T p.M40L Chr.3: 49747923-49747923 on Build GRCh38
1617767 c.108C>T p.H36H Chr.3: 49747933-49747933 on Build GRCh38
6686581 c.151delC p.L51fs*16 Chr.3: 49747890-49747890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055937877 Chr.3: 49748008-49748008 on Build GRCh38
rs747206201 Chr.3: 49747992-49747992 on Build GRCh38
rs891799918 Chr.3: 49747988-49747988 on Build GRCh38
rs762695026 Chr.3: 49748018-49748018 on Build GRCh38
rs765008760 Chr.3: 49747918-49747918 on Build GRCh38
rs758652211 Chr.3: 49748026-49748026 on Build GRCh38
rs772467381 Chr.3: 49747893-49747893 on Build GRCh38
rs780286190 Chr.3: 49747991-49747991 on Build GRCh38
rs370834861 Chr.3: 49747989-49747989 on Build GRCh38
rs187248712 Chr.3: 49747882-49747882 on Build GRCh38
rs201282630 Chr.3: 49747976-49747976 on Build GRCh38
rs748043612 Chr.3: 49748004-49748004 on Build GRCh38
rs966549850 Chr.3: 49748006-49748006 on Build GRCh38
rs752296772 Chr.3: 49748067-49748067 on Build GRCh38
rs34785187 Chr.3: 49747997-49747997 on Build GRCh38
rs750257907 Chr.3: 49747919-49747919 on Build GRCh38
rs541583110 Chr.3: 49747969-49747969 on Build GRCh38
rs760761629 Chr.3: 49747909-49747909 on Build GRCh38
rs139564570 Chr.3: 49748007-49748007 on Build GRCh38
rs765794820 Chr.3: 49748077-49748077 on Build GRCh38
rs754445993 Chr.3: 49747953-49747953 on Build GRCh38
rs146101769 Chr.3: 49748014-49748014 on Build GRCh38
rs759170952 Chr.3: 49748051-49748051 on Build GRCh38
rs553293311 Chr.3: 49747915-49747915 on Build GRCh38
rs112398797 Chr.3: 49747926-49747926 on Build GRCh38
rs768765320 Chr.3: 49747996-49747996 on Build GRCh38
rs779393912 Chr.3: 49747986-49747986 on Build GRCh38
rs377655659 Chr.3: 49747880-49747880 on Build GRCh38
rs773998985 Chr.3: 49748027-49748027 on Build GRCh38
rs766952095 Chr.3: 49747954-49747954 on Build GRCh38
rs768653218 Chr.3: 49747912-49747912 on Build GRCh38
rs775413636 Chr.3: 49747897-49747897 on Build GRCh38
rs953887313 Chr.3: 49748075-49748075 on Build GRCh38
rs751200749 Chr.3: 49747951-49747951 on Build GRCh38
rs750517104 Chr.3: 49748052-49748052 on Build GRCh38
rs767049966 Chr.3: 49748064-49748064 on Build GRCh38
rs779478732 Chr.3: 49747881-49747881 on Build GRCh38
rs776571630 Chr.3: 49747914-49747914 on Build GRCh38
rs766006261 Chr.3: 49747921-49747921 on Build GRCh38
rs1028543445 Chr.3: 49748044-49748044 on Build GRCh38
rs1014370740 Chr.3: 49748059-49748059 on Build GRCh38
rs757819905 Chr.3: 49748069-49748069 on Build GRCh38

View Full Product Details