Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 28767820-28768085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXG1
Entrez Gene ID
2290
Gene Name
forkhead box G1
Gene Aliases
BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN
UniGene
Hs.632336
Gene Chromosome Location
Chr.14: 28767072-28770277 on Build GRCh38
Transcripts
NM_005249.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3690052 c.724G>A p.V242M Chr.14: 28768003-28768003 on Build GRCh38
2033899 c.802G>A p.G268S Chr.14: 28768081-28768081 on Build GRCh38
2033896 c.731G>A p.R244H Chr.14: 28768010-28768010 on Build GRCh38
2033894 c.696T>C p.N232N Chr.14: 28767975-28767975 on Build GRCh38
2153502 c.685_686delAT p.I229fs*225 Chr.14: 28767964-28767965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759790790 Chr.14: 28768029-28768029 on Build GRCh38
rs768525699 Chr.14: 28767936-28767936 on Build GRCh38
rs786205005 Chr.14: 28767856-28767856 on Build GRCh38
rs796052468 Chr.14: 28767987-28767987 on Build GRCh38
rs587783639 Chr.14: 28767951-28767951 on Build GRCh38
rs121913678 Chr.14: 28768044-28768044 on Build GRCh38
rs267606828 Chr.14: 28767922-28767922 on Build GRCh38
rs796052481 Chr.14: 28767950-28767950 on Build GRCh38
rs869312961 Chr.14: 28768055-28768055 on Build GRCh38
rs930327703 Chr.14: 28767897-28767897 on Build GRCh38
rs756719183 Chr.14: 28767861-28767861 on Build GRCh38
rs796052484 Chr.14: 28768010-28768010 on Build GRCh38
rs786205009 Chr.14: 28768009-28768009 on Build GRCh38
rs1057518165 Chr.14: 28767924-28767924 on Build GRCh38
rs796052483 Chr.14: 28768000-28768000 on Build GRCh38
rs587783640 Chr.14: 28768034-28768034 on Build GRCh38
rs771755778 Chr.14: 28767900-28767900 on Build GRCh38
rs796052466 Chr.14: 28767954-28767954 on Build GRCh38
rs786205008 Chr.14: 28767979-28767979 on Build GRCh38
rs796052462 Chr.14: 28767840-28767840 on Build GRCh38
rs796052465 Chr.14: 28767946-28767946 on Build GRCh38
rs786205486 Chr.14: 28767973-28767973 on Build GRCh38
rs199502880 Chr.14: 28767965-28767965 on Build GRCh38
rs587783641 Chr.14: 28768036-28768036 on Build GRCh38
rs796052463 Chr.14: 28767865-28767865 on Build GRCh38
rs766343459 Chr.14: 28767969-28767969 on Build GRCh38
rs774525510 Chr.14: 28768008-28768008 on Build GRCh38
rs905973556 Chr.14: 28767849-28767849 on Build GRCh38
rs141088742 Chr.14: 28767873-28767873 on Build GRCh38
rs748766934 Chr.14: 28767846-28767846 on Build GRCh38
rs727503935 Chr.14: 28767949-28767949 on Build GRCh38
rs1001642335 Chr.14: 28767863-28767863 on Build GRCh38
rs796052467 Chr.14: 28767980-28767980 on Build GRCh38
rs796052482 Chr.14: 28767952-28767952 on Build GRCh38
rs267606826 Chr.14: 28767903-28767903 on Build GRCh38
rs201191801 Chr.14: 28767885-28767885 on Build GRCh38
rs786205010 Chr.14: 28768067-28768071 on Build GRCh38
rs786205006 Chr.14: 28767889-28767889 on Build GRCh38
rs786205012 Chr.14: 28767960-28767960 on Build GRCh38
rs796934848 Chr.14: 28768062-28768062 on Build GRCh38
rs786204998 Chr.14: 28767923-28767924 on Build GRCh38
rs786205007 Chr.14: 28767968-28767968 on Build GRCh38
rs767873754 Chr.14: 28768038-28768038 on Build GRCh38
rs727503934 Chr.14: 28767927-28767934 on Build GRCh38
rs761866084 Chr.14: 28767963-28767963 on Build GRCh38
rs587783642 Chr.14: 28768041-28768041 on Build GRCh38
rs587783638 Chr.14: 28767842-28767842 on Build GRCh38
rs796052464 Chr.14: 28767930-28767930 on Build GRCh38
rs960034268 Chr.14: 28768068-28768068 on Build GRCh38
rs745486814 Chr.14: 28767880-28767880 on Build GRCh38
rs548935590 Chr.14: 28767925-28767925 on Build GRCh38

View Full Product Details