Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1804370-1804641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1
Entrez Gene ID
2782
Gene Name
G protein subunit beta 1
Gene Aliases
MRD42
UniGene
Hs.430425
Gene Chromosome Location
Chr.1: 1785285-1891117 on Build GRCh38
Transcripts
NM_001282538.1 NM_001282539.1 NM_002074.4 XM_017001059.1 XM_017001060.1 XM_017001061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4748560 c.403G>A p.V135M Chr.1: 1804446-1804446 on Build GRCh38
6672271 c.343G>A p.G115S Chr.1: 1804506-1804506 on Build GRCh38
2075607 c.314A>G p.Y105C Chr.1: 1804535-1804535 on Build GRCh38
1336398 c.287G>A p.R96H Chr.1: 1804562-1804562 on Build GRCh38
4025211 c.402C>T p.R134R Chr.1: 1804447-1804447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747251185 Chr.1: 1804591-1804592 on Build GRCh38
rs746909946 Chr.1: 1804555-1804555 on Build GRCh38
rs940163168 Chr.1: 1804450-1804450 on Build GRCh38
rs781404706 Chr.1: 1804452-1804452 on Build GRCh38
rs764585029 Chr.1: 1804415-1804415 on Build GRCh38
rs764897408 Chr.1: 1804593-1804593 on Build GRCh38
rs201240857 Chr.1: 1804592-1804592 on Build GRCh38
rs202060609 Chr.1: 1804395-1804395 on Build GRCh38
rs751949845 Chr.1: 1804426-1804426 on Build GRCh38
rs757859923 Chr.1: 1804410-1804410 on Build GRCh38
rs869312824 Chr.1: 1804565-1804565 on Build GRCh38
rs764770433 Chr.1: 1804402-1804402 on Build GRCh38
rs373455868 Chr.1: 1804594-1804594 on Build GRCh38
rs369599792 Chr.1: 1804405-1804405 on Build GRCh38
rs888774835 Chr.1: 1804537-1804537 on Build GRCh38
rs869312825 Chr.1: 1804548-1804548 on Build GRCh38
rs1009584158 Chr.1: 1804400-1804400 on Build GRCh38
rs1036128394 Chr.1: 1804471-1804471 on Build GRCh38
rs897548786 Chr.1: 1804472-1804472 on Build GRCh38
rs112027616 Chr.1: 1804416-1804416 on Build GRCh38
rs150628410 Chr.1: 1804475-1804475 on Build GRCh38
rs376445565 Chr.1: 1804602-1804602 on Build GRCh38
rs749411068 Chr.1: 1804516-1804516 on Build GRCh38
rs763496799 Chr.1: 1804613-1804613 on Build GRCh38
rs368984912 Chr.1: 1804486-1804486 on Build GRCh38
rs780373436 Chr.1: 1804510-1804510 on Build GRCh38
rs200842912 Chr.1: 1804549-1804549 on Build GRCh38
rs556077872 Chr.1: 1804507-1804507 on Build GRCh38
rs560214051 Chr.1: 1804447-1804447 on Build GRCh38
rs768790682 Chr.1: 1804531-1804531 on Build GRCh38
rs776262309 Chr.1: 1804576-1804576 on Build GRCh38
rs773542848 Chr.1: 1804587-1804588 on Build GRCh38
rs775053997 Chr.1: 1804601-1804601 on Build GRCh38
rs770828889 Chr.1: 1804561-1804561 on Build GRCh38

View Full Product Details