Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37056275-37056508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD17
Entrez Gene ID
79734
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
-
UniGene
Hs.517597
Gene Chromosome Location
Chr.22: 37051725-37063390 on Build GRCh38
Transcripts
NM_001282684.1 NM_001282685.1 NM_001282686.1 NM_024681.3 XM_005261741.3 XM_005261742.2 XM_005261743.2 XM_005261744.2 XM_011530374.2 XM_011530377.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692053 c.401C>T p.T134M Chr.22: 37056401-37056401 on Build GRCh38
4753005 c.380G>A p.R127Q Chr.22: 37056380-37056380 on Build GRCh38
4801673 c.336C>A p.A112A Chr.22: 37056336-37056336 on Build GRCh38
4801672 c.315C>A p.A105A Chr.22: 37056336-37056336 on Build GRCh38
7081641 c.405C>T p.V135V Chr.22: 37056405-37056405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745889631 Chr.22: 37056372-37056372 on Build GRCh38
rs894583805 Chr.22: 37056307-37056307 on Build GRCh38
rs1056185601 Chr.22: 37056304-37056304 on Build GRCh38
rs977261880 Chr.22: 37056479-37056479 on Build GRCh38
rs746122376 Chr.22: 37056454-37056454 on Build GRCh38
rs781117547 Chr.22: 37056367-37056367 on Build GRCh38
rs777509126 Chr.22: 37056477-37056477 on Build GRCh38
rs372826685 Chr.22: 37056351-37056351 on Build GRCh38
rs150904187 Chr.22: 37056336-37056336 on Build GRCh38
rs749246602 Chr.22: 37056386-37056386 on Build GRCh38
rs747215623 Chr.22: 37056412-37056412 on Build GRCh38
rs781762947 Chr.22: 37056458-37056459 on Build GRCh38
rs781410115 Chr.22: 37056447-37056447 on Build GRCh38
rs532452879 Chr.22: 37056424-37056424 on Build GRCh38
rs760219436 Chr.22: 37056415-37056415 on Build GRCh38
rs750023360 Chr.22: 37056320-37056320 on Build GRCh38
rs769868923 Chr.22: 37056380-37056380 on Build GRCh38
rs887307685 Chr.22: 37056337-37056337 on Build GRCh38
rs149432408 Chr.22: 37056417-37056417 on Build GRCh38
rs773015272 Chr.22: 37056414-37056414 on Build GRCh38
rs188116230 Chr.22: 37056482-37056482 on Build GRCh38
rs11542756 Chr.22: 37056394-37056394 on Build GRCh38
rs955319291 Chr.22: 37056385-37056385 on Build GRCh38
rs747167411 Chr.22: 37056364-37056364 on Build GRCh38
rs774245782 Chr.22: 37056402-37056402 on Build GRCh38
rs183791733 Chr.22: 37056435-37056435 on Build GRCh38
rs757565419 Chr.22: 37056365-37056365 on Build GRCh38
rs370855894 Chr.22: 37056401-37056401 on Build GRCh38
rs761372052 Chr.22: 37056407-37056407 on Build GRCh38
rs35901190 Chr.22: 37056357-37056357 on Build GRCh38
rs775369238 Chr.22: 37056382-37056382 on Build GRCh38
rs371921045 Chr.22: 37056436-37056436 on Build GRCh38
rs536277554 Chr.22: 37056422-37056422 on Build GRCh38
rs755532238 Chr.22: 37056327-37056327 on Build GRCh38
rs1031746578 Chr.22: 37056408-37056408 on Build GRCh38
rs369091372 Chr.22: 37056440-37056440 on Build GRCh38
rs758448853 Chr.22: 37056332-37056332 on Build GRCh38
rs766909394 Chr.22: 37056315-37056315 on Build GRCh38
rs764361408 Chr.22: 37056427-37056427 on Build GRCh38
rs76442271 Chr.22: 37056352-37056352 on Build GRCh38
rs533725861 Chr.22: 37056474-37056474 on Build GRCh38

View Full Product Details