Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41281090-41281363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707033 c.949G>A p.E317K Chr.19: 41281225-41281225 on Build GRCh38
1750920 c.887-1G>T p.? Chr.19: 41281162-41281162 on Build GRCh38
996993 c.888C>T p.G296G Chr.19: 41281164-41281164 on Build GRCh38
1750921 c.958G>T p.G320* Chr.19: 41281234-41281234 on Build GRCh38
996994 c.958G>A p.G320R Chr.19: 41281234-41281234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752760597 Chr.19: 41281215-41281215 on Build GRCh38
rs774145913 Chr.19: 41281316-41281316 on Build GRCh38
rs555562080 Chr.19: 41281191-41281191 on Build GRCh38
rs768244832 Chr.19: 41281157-41281157 on Build GRCh38
rs780709153 Chr.19: 41281274-41281274 on Build GRCh38
rs767618709 Chr.19: 41281113-41281115 on Build GRCh38
rs774052802 Chr.19: 41281208-41281208 on Build GRCh38
rs994422263 Chr.19: 41281312-41281312 on Build GRCh38
rs142945712 Chr.19: 41281247-41281247 on Build GRCh38
rs747685510 Chr.19: 41281176-41281176 on Build GRCh38
rs764327881 Chr.19: 41281220-41281220 on Build GRCh38
rs377049768 Chr.19: 41281118-41281118 on Build GRCh38
rs769275998 Chr.19: 41281292-41281292 on Build GRCh38
rs778431167 Chr.19: 41281126-41281126 on Build GRCh38
rs777331612 Chr.19: 41281293-41281293 on Build GRCh38
rs191480843 Chr.19: 41281125-41281125 on Build GRCh38
rs779317116 Chr.19: 41281240-41281240 on Build GRCh38
rs572276508 Chr.19: 41281239-41281239 on Build GRCh38
rs543996226 Chr.19: 41281315-41281315 on Build GRCh38
rs749607501 Chr.19: 41281129-41281129 on Build GRCh38
rs757682940 Chr.19: 41281136-41281136 on Build GRCh38
rs200985635 Chr.19: 41281214-41281214 on Build GRCh38
rs760852808 Chr.19: 41281219-41281219 on Build GRCh38
rs375997855 Chr.19: 41281289-41281289 on Build GRCh38
rs746645347 Chr.19: 41281156-41281156 on Build GRCh38
rs74529580 Chr.19: 41281273-41281273 on Build GRCh38
rs754392087 Chr.19: 41281227-41281227 on Build GRCh38
rs755222103 Chr.19: 41281117-41281117 on Build GRCh38
rs759256905 Chr.19: 41281323-41281323 on Build GRCh38
rs916899152 Chr.19: 41281170-41281170 on Build GRCh38
rs748633563 Chr.19: 41281164-41281164 on Build GRCh38
rs540686842 Chr.19: 41281182-41281182 on Build GRCh38
rs757748656 Chr.19: 41281238-41281238 on Build GRCh38
rs750778968 Chr.19: 41281254-41281254 on Build GRCh38
rs754621449 Chr.19: 41281263-41281263 on Build GRCh38
rs749191568 Chr.19: 41281303-41281303 on Build GRCh38
rs562455909 Chr.19: 41281325-41281325 on Build GRCh38
rs759779569 Chr.19: 41281210-41281210 on Build GRCh38
rs769649013 Chr.19: 41281179-41281179 on Build GRCh38
rs771660545 Chr.19: 41281288-41281288 on Build GRCh38
rs368451935 Chr.19: 41281149-41281149 on Build GRCh38
rs1027545124 Chr.19: 41281337-41281337 on Build GRCh38
rs780707292 Chr.19: 41281158-41281158 on Build GRCh38

View Full Product Details