Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55107655-55107884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMA
Entrez Gene ID
3001
Gene Name
granzyme A
Gene Aliases
CTLA3, HFSP
UniGene
Hs.90708
Gene Chromosome Location
Chr.5: 55102646-55110252 on Build GRCh38
Transcripts
NM_006144.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5001919 c.304C>A p.P102T Chr.5: 55107882-55107882 on Build GRCh38
385183 c.258C>A p.T86T Chr.5: 55107836-55107836 on Build GRCh38
173043 c.238C>A p.L80I Chr.5: 55107816-55107816 on Build GRCh38
3617014 c.239T>C p.L80P Chr.5: 55107817-55107817 on Build GRCh38
1253936 c.276A>G p.K92K Chr.5: 55107854-55107854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770229169 Chr.5: 55107851-55107851 on Build GRCh38
rs752702531 Chr.5: 55107810-55107810 on Build GRCh38
rs760413719 Chr.5: 55107752-55107752 on Build GRCh38
rs760176125 Chr.5: 55107829-55107829 on Build GRCh38
rs200389236 Chr.5: 55107760-55107760 on Build GRCh38
rs180863855 Chr.5: 55107751-55107751 on Build GRCh38
rs370970693 Chr.5: 55107851-55107851 on Build GRCh38
rs764546928 Chr.5: 55107748-55107748 on Build GRCh38
rs756002359 Chr.5: 55107678-55107678 on Build GRCh38
rs754934377 Chr.5: 55107776-55107776 on Build GRCh38
rs923091205 Chr.5: 55107820-55107820 on Build GRCh38
rs765972102 Chr.5: 55107758-55107758 on Build GRCh38
rs989785369 Chr.5: 55107724-55107724 on Build GRCh38
rs914265631 Chr.5: 55107733-55107733 on Build GRCh38
rs758063250 Chr.5: 55107825-55107825 on Build GRCh38
rs936966281 Chr.5: 55107716-55107716 on Build GRCh38
rs763183459 Chr.5: 55107747-55107747 on Build GRCh38
rs141463827 Chr.5: 55107850-55107850 on Build GRCh38
rs139073733 Chr.5: 55107843-55107843 on Build GRCh38
rs765136087 Chr.5: 55107804-55107804 on Build GRCh38
rs267600651 Chr.5: 55107840-55107840 on Build GRCh38
rs774869229 Chr.5: 55107776-55107778 on Build GRCh38
rs1030683001 Chr.5: 55107738-55107738 on Build GRCh38

View Full Product Details