Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146515506-146515779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR151
Entrez Gene ID
134391
Gene Name
G protein-coupled receptor 151
Gene Aliases
GALR4, GALRL, GPCR, PGR7
UniGene
Hs.483732
Gene Chromosome Location
Chr.5: 146514854-146516113 on Build GRCh38
Transcripts
NM_194251.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5756590 c.494C>A p.A165D Chr.5: 146515620-146515620 on Build GRCh38
3852436 c.513G>A p.P171P Chr.5: 146515601-146515601 on Build GRCh38
5906754 c.486G>A p.W162* Chr.5: 146515628-146515628 on Build GRCh38
3210331 c.568G>A p.D190N Chr.5: 146515546-146515546 on Build GRCh38
7146602 c.467C>T p.S156L Chr.5: 146515647-146515647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202188079 Chr.5: 146515750-146515750 on Build GRCh38
rs750801084 Chr.5: 146515603-146515604 on Build GRCh38
rs138413807 Chr.5: 146515686-146515686 on Build GRCh38
rs764829766 Chr.5: 146515609-146515609 on Build GRCh38
rs780560800 Chr.5: 146515722-146515724 on Build GRCh38
rs369802204 Chr.5: 146515726-146515726 on Build GRCh38
rs780486421 Chr.5: 146515537-146515537 on Build GRCh38
rs267600472 Chr.5: 146515747-146515747 on Build GRCh38
rs144403542 Chr.5: 146515696-146515696 on Build GRCh38
rs758113148 Chr.5: 146515668-146515668 on Build GRCh38
rs149603566 Chr.5: 146515687-146515687 on Build GRCh38
rs753234875 Chr.5: 146515749-146515749 on Build GRCh38
rs751873529 Chr.5: 146515532-146515536 on Build GRCh38
rs183701313 Chr.5: 146515549-146515549 on Build GRCh38
rs768441360 Chr.5: 146515697-146515697 on Build GRCh38
rs894028619 Chr.5: 146515646-146515646 on Build GRCh38
rs762221581 Chr.5: 146515539-146515542 on Build GRCh38
rs752031026 Chr.5: 146515676-146515676 on Build GRCh38
rs761827694 Chr.5: 146515701-146515701 on Build GRCh38
rs368648211 Chr.5: 146515755-146515755 on Build GRCh38
rs551340129 Chr.5: 146515753-146515753 on Build GRCh38
rs759941471 Chr.5: 146515744-146515744 on Build GRCh38
rs772849621 Chr.5: 146515714-146515714 on Build GRCh38
rs771741753 Chr.5: 146515566-146515566 on Build GRCh38
rs758646176 Chr.5: 146515534-146515534 on Build GRCh38
rs751264556 Chr.5: 146515741-146515741 on Build GRCh38
rs755410753 Chr.5: 146515680-146515680 on Build GRCh38
rs775304062 Chr.5: 146515570-146515570 on Build GRCh38
rs766073591 Chr.5: 146515727-146515727 on Build GRCh38
rs371947982 Chr.5: 146515552-146515552 on Build GRCh38
rs747492655 Chr.5: 146515539-146515539 on Build GRCh38
rs111754680 Chr.5: 146515601-146515601 on Build GRCh38
rs7713676 Chr.5: 146515683-146515683 on Build GRCh38
rs755612829 Chr.5: 146515540-146515540 on Build GRCh38
rs766652832 Chr.5: 146515674-146515674 on Build GRCh38
rs756533726 Chr.5: 146515674-146515677 on Build GRCh38
rs760600394 Chr.5: 146515571-146515571 on Build GRCh38
rs139626065 Chr.5: 146515707-146515707 on Build GRCh38
rs750091501 Chr.5: 146515533-146515533 on Build GRCh38
rs748183557 Chr.5: 146515550-146515550 on Build GRCh38
rs908021456 Chr.5: 146515721-146515721 on Build GRCh38
rs1039317557 Chr.5: 146515617-146515617 on Build GRCh38
rs529442982 Chr.5: 146515700-146515700 on Build GRCh38
rs929400387 Chr.5: 146515577-146515577 on Build GRCh38
rs764075409 Chr.5: 146515572-146515572 on Build GRCh38
rs375295268 Chr.5: 146515684-146515684 on Build GRCh38
rs775250424 Chr.5: 146515694-146515694 on Build GRCh38
rs558001563 Chr.5: 146515588-146515589 on Build GRCh38
rs902294436 Chr.5: 146515706-146515706 on Build GRCh38
rs749439222 Chr.5: 146515688-146515688 on Build GRCh38
rs1035122198 Chr.5: 146515718-146515718 on Build GRCh38
rs943343718 Chr.5: 146515613-146515613 on Build GRCh38
rs769923462 Chr.5: 146515551-146515551 on Build GRCh38
rs985333845 Chr.5: 146515719-146515719 on Build GRCh38
rs771251688 Chr.5: 146515692-146515692 on Build GRCh38
rs551697657 Chr.5: 146515616-146515616 on Build GRCh38
rs749548966 Chr.5: 146515559-146515559 on Build GRCh38
rs776560462 Chr.5: 146515593-146515593 on Build GRCh38
rs148792036 Chr.5: 146515699-146515699 on Build GRCh38
rs772502654 Chr.5: 146515663-146515663 on Build GRCh38

View Full Product Details