Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29604667-29604902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABBR1
Entrez Gene ID
2550
Gene Name
gamma-aminobutyric acid type B receptor subunit 1
Gene Aliases
GABABR1, GABBR1-3, GB1, GPRC3A
UniGene
Hs.167017
Gene Chromosome Location
Chr.6: 29602228-29633135 on Build GRCh38
Transcripts
NM_001319053.1 NM_001470.3 NM_021903.2 NM_021904.3 XM_005248982.2 XM_006715047.3 XM_011514453.2 XM_017010676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5678613 c.2217G>A p.K739K Chr.6: 29604860-29604860 on Build GRCh38
5678612 c.2568G>A p.K856K Chr.6: 29604860-29604860 on Build GRCh38
7183846 c.2556C>G p.L852L Chr.6: 29604872-29604872 on Build GRCh38
7183847 c.2205C>G p.L735L Chr.6: 29604872-29604872 on Build GRCh38
4686068 c.2561T>C p.V854A Chr.6: 29604867-29604867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565465963 Chr.6: 29604730-29604730 on Build GRCh38
rs190005485 Chr.6: 29604710-29604710 on Build GRCh38
rs1053794019 Chr.6: 29604824-29604824 on Build GRCh38
rs369743030 Chr.6: 29604866-29604866 on Build GRCh38
rs561568814 Chr.6: 29604796-29604796 on Build GRCh38
rs903540276 Chr.6: 29604835-29604835 on Build GRCh38
rs545914368 Chr.6: 29604713-29604713 on Build GRCh38
rs377713883 Chr.6: 29604748-29604748 on Build GRCh38
rs112562687 Chr.6: 29604688-29604688 on Build GRCh38
rs762299868 Chr.6: 29604816-29604816 on Build GRCh38
rs768069286 Chr.6: 29604822-29604822 on Build GRCh38
rs181702336 Chr.6: 29604784-29604784 on Build GRCh38
rs531172373 Chr.6: 29604772-29604772 on Build GRCh38
rs553796810 Chr.6: 29604704-29604704 on Build GRCh38
rs760847538 Chr.6: 29604855-29604855 on Build GRCh38
rs917107765 Chr.6: 29604705-29604705 on Build GRCh38
rs977783187 Chr.6: 29604763-29604763 on Build GRCh38
rs530061014 Chr.6: 29604869-29604869 on Build GRCh38
rs916076733 Chr.6: 29604773-29604773 on Build GRCh38
rs750478180 Chr.6: 29604840-29604840 on Build GRCh38

View Full Product Details