Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156623386-156623659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN2
Entrez Gene ID
60484
Gene Name
hyaluronan and proteoglycan link protein 2
Gene Aliases
BRAL1
UniGene
Hs.410719
Gene Chromosome Location
Chr.1: 156601504-156626608 on Build GRCh38
Transcripts
NM_021817.2 XM_005245415.2 XM_011509853.2 XM_017002020.1 XM_017002021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941662 c.84A>G p.P28P Chr.1: 156623574-156623574 on Build GRCh38
140381 c.40C>T p.L14F Chr.1: 156623530-156623530 on Build GRCh38
1983882 c.61T>A p.F21I Chr.1: 156623551-156623551 on Build GRCh38
897275 c.70G>A p.A24T Chr.1: 156623560-156623560 on Build GRCh38
7290391 c.51C>A p.A17A Chr.1: 156623541-156623541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988403397 Chr.1: 156623565-156623565 on Build GRCh38
rs770247265 Chr.1: 156623612-156623612 on Build GRCh38
rs773410470 Chr.1: 156623509-156623517 on Build GRCh38
rs200755008 Chr.1: 156623468-156623468 on Build GRCh38
rs748779498 Chr.1: 156623481-156623482 on Build GRCh38
rs777327763 Chr.1: 156623479-156623479 on Build GRCh38
rs772377323 Chr.1: 156623472-156623472 on Build GRCh38
rs748025834 Chr.1: 156623456-156623456 on Build GRCh38
rs752334491 Chr.1: 156623453-156623454 on Build GRCh38
rs765822097 Chr.1: 156623481-156623481 on Build GRCh38
rs755622129 Chr.1: 156623464-156623464 on Build GRCh38
rs1054639731 Chr.1: 156623452-156623452 on Build GRCh38
rs776551066 Chr.1: 156623607-156623607 on Build GRCh38
rs754721614 Chr.1: 156623524-156623524 on Build GRCh38
rs775857707 Chr.1: 156623473-156623473 on Build GRCh38
rs200487120 Chr.1: 156623505-156623505 on Build GRCh38
rs755954336 Chr.1: 156623545-156623545 on Build GRCh38
rs762196013 Chr.1: 156623480-156623480 on Build GRCh38
rs907135878 Chr.1: 156623602-156623602 on Build GRCh38
rs768963591 Chr.1: 156623606-156623606 on Build GRCh38
rs757234514 Chr.1: 156623497-156623500 on Build GRCh38
rs752295940 Chr.1: 156623540-156623540 on Build GRCh38
rs946529216 Chr.1: 156623640-156623640 on Build GRCh38
rs150142304 Chr.1: 156623489-156623489 on Build GRCh38
rs777880728 Chr.1: 156623548-156623548 on Build GRCh38
rs896064099 Chr.1: 156623409-156623409 on Build GRCh38
rs1025795702 Chr.1: 156623432-156623432 on Build GRCh38
rs373807773 Chr.1: 156623557-156623557 on Build GRCh38
rs368705677 Chr.1: 156623451-156623451 on Build GRCh38
rs747190633 Chr.1: 156623591-156623591 on Build GRCh38
rs138640704 Chr.1: 156623562-156623562 on Build GRCh38
rs534110586 Chr.1: 156623415-156623415 on Build GRCh38
rs558442010 Chr.1: 156623445-156623445 on Build GRCh38
rs778909567 Chr.1: 156623585-156623585 on Build GRCh38
rs1014878991 Chr.1: 156623418-156623418 on Build GRCh38
rs375772454 Chr.1: 156623475-156623475 on Build GRCh38
rs748284267 Chr.1: 156623611-156623611 on Build GRCh38
rs964040626 Chr.1: 156623513-156623513 on Build GRCh38
rs369091064 Chr.1: 156623622-156623622 on Build GRCh38
rs766935922 Chr.1: 156623525-156623525 on Build GRCh38
rs913748585 Chr.1: 156623629-156623629 on Build GRCh38
rs773518242 Chr.1: 156623487-156623487 on Build GRCh38
rs537533829 Chr.1: 156623469-156623469 on Build GRCh38
rs779572483 Chr.1: 156623503-156623503 on Build GRCh38

View Full Product Details