Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73111263-73111494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ5
Entrez Gene ID
56479
Gene Name
potassium voltage-gated channel subfamily Q member 5
Gene Aliases
Kv7.5
UniGene
Hs.445324
Gene Chromosome Location
Chr.6: 72621843-73198851 on Build GRCh38
Transcripts
NM_001160130.1 NM_001160132.1 NM_001160133.1 NM_001160134.1 NM_019842.3 XM_017011058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3922126 c.1097G>A p.R366K Chr.6: 73111375-73111375 on Build GRCh38
7076323 c.1106C>T p.P369L Chr.6: 73111384-73111384 on Build GRCh38
5871077 c.1046G>A p.G349D Chr.6: 73111324-73111324 on Build GRCh38
7076324 c.1106C>T p.P369L Chr.6: 73111384-73111384 on Build GRCh38
7076325 c.1106C>T p.P369L Chr.6: 73111384-73111384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765290049 Chr.6: 73111410-73111410 on Build GRCh38
rs775342463 Chr.6: 73111301-73111301 on Build GRCh38
rs761119223 Chr.6: 73111319-73111319 on Build GRCh38
rs764408697 Chr.6: 73111331-73111331 on Build GRCh38
rs755915115 Chr.6: 73111450-73111450 on Build GRCh38
rs745448246 Chr.6: 73111291-73111291 on Build GRCh38
rs758528734 Chr.6: 73111420-73111420 on Build GRCh38
rs762253123 Chr.6: 73111382-73111382 on Build GRCh38
rs149249368 Chr.6: 73111376-73111376 on Build GRCh38
rs1034900230 Chr.6: 73111421-73111421 on Build GRCh38
rs771865627 Chr.6: 73111294-73111294 on Build GRCh38
rs766582582 Chr.6: 73111445-73111445 on Build GRCh38
rs1935538 Chr.6: 73111446-73111446 on Build GRCh38
rs45536838 Chr.6: 73111290-73111290 on Build GRCh38
rs1935537 Chr.6: 73111470-73111470 on Build GRCh38

View Full Product Details