Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113820823-113821062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS6
Entrez Gene ID
2621
Gene Name
growth arrest specific 6
Gene Aliases
AXLLG, AXSF
UniGene
Hs.646346
Gene Chromosome Location
Chr.13: 113820549-113864103 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Gene Symbol
GAS6-AS1
Entrez Gene ID
650669
Gene Name
GAS6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.369201
Gene Chromosome Location
Chr.13: 113815610-113842841 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2264537 c.2016delC p.V673fs*>6 Chr.13: 113820885-113820885 on Build GRCh38
6669444 c.1855G>A p.V619M Chr.13: 113820884-113820884 on Build GRCh38
7181520 c.1930A>G p.M644V Chr.13: 113820971-113820971 on Build GRCh38
4786518 c.1818C>T p.Y606Y Chr.13: 113820921-113820921 on Build GRCh38
4785115 c.2160A>G p.A720A Chr.13: 113820870-113820870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775825887 Chr.13: 113820960-113820960 on Build GRCh38
rs751304956 Chr.13: 113821027-113821028 on Build GRCh38
rs201330919 Chr.13: 113820932-113820932 on Build GRCh38
rs770666266 Chr.13: 113820933-113820933 on Build GRCh38
rs780686636 Chr.13: 113820993-113820993 on Build GRCh38
rs141047065 Chr.13: 113821021-113821021 on Build GRCh38
rs776020399 Chr.13: 113820876-113820876 on Build GRCh38
rs750258083 Chr.13: 113821026-113821026 on Build GRCh38
rs77469272 Chr.13: 113820883-113820883 on Build GRCh38
rs777011915 Chr.13: 113820857-113820858 on Build GRCh38
rs768559329 Chr.13: 113821037-113821037 on Build GRCh38
rs754684052 Chr.13: 113820924-113820924 on Build GRCh38
rs143026609 Chr.13: 113820997-113820997 on Build GRCh38
rs77167214 Chr.13: 113820885-113820885 on Build GRCh38
rs371109462 Chr.13: 113821031-113821031 on Build GRCh38
rs1008695454 Chr.13: 113820928-113820928 on Build GRCh38
rs754407477 Chr.13: 113820908-113820908 on Build GRCh38
rs766199351 Chr.13: 113820885-113820885 on Build GRCh38
rs748053207 Chr.13: 113820878-113820878 on Build GRCh38
rs1044549596 Chr.13: 113821032-113821032 on Build GRCh38
rs772168900 Chr.13: 113820953-113820953 on Build GRCh38
rs555027211 Chr.13: 113820848-113820848 on Build GRCh38
rs764264848 Chr.13: 113820982-113820982 on Build GRCh38
rs767619005 Chr.13: 113820849-113820849 on Build GRCh38
rs140807230 Chr.13: 113820927-113820927 on Build GRCh38
rs765629353 Chr.13: 113820988-113820988 on Build GRCh38
rs77565607 Chr.13: 113820884-113820884 on Build GRCh38
rs999300128 Chr.13: 113820855-113820855 on Build GRCh38
rs750814570 Chr.13: 113820989-113820989 on Build GRCh38
rs779822614 Chr.13: 113820872-113820872 on Build GRCh38
rs147032954 Chr.13: 113821012-113821012 on Build GRCh38
rs200546417 Chr.13: 113820952-113820952 on Build GRCh38
rs893288560 Chr.13: 113820862-113820862 on Build GRCh38
rs911109356 Chr.13: 113820967-113820967 on Build GRCh38
rs760636776 Chr.13: 113820978-113820978 on Build GRCh38
rs757741900 Chr.13: 113820864-113820864 on Build GRCh38
rs759579441 Chr.13: 113821036-113821038 on Build GRCh38
rs747147158 Chr.13: 113820971-113820971 on Build GRCh38
rs752712382 Chr.13: 113820892-113820892 on Build GRCh38
rs769396670 Chr.13: 113820926-113820926 on Build GRCh38
rs780966895 Chr.13: 113820925-113820925 on Build GRCh38
rs760861912 Chr.13: 113820903-113820903 on Build GRCh38
rs768231297 Chr.13: 113820875-113820875 on Build GRCh38
rs756580750 Chr.13: 113820851-113820851 on Build GRCh38
rs372674142 Chr.13: 113821000-113821000 on Build GRCh38
rs893585319 Chr.13: 113821027-113821027 on Build GRCh38
rs753201452 Chr.13: 113820850-113820850 on Build GRCh38
rs958437135 Chr.13: 113821024-113821024 on Build GRCh38
rs1036128775 Chr.13: 113820980-113820980 on Build GRCh38
rs76238089 Chr.13: 113820860-113820860 on Build GRCh38
rs774149044 Chr.13: 113820945-113820945 on Build GRCh38
rs150170581 Chr.13: 113820987-113820987 on Build GRCh38
rs144441512 Chr.13: 113820912-113820912 on Build GRCh38
rs775534088 Chr.13: 113821029-113821029 on Build GRCh38
rs371833945 Chr.13: 113820884-113820885 on Build GRCh38
rs755543652 Chr.13: 113820999-113820999 on Build GRCh38
rs544848038 Chr.13: 113820891-113820891 on Build GRCh38
rs749169005 Chr.13: 113821011-113821011 on Build GRCh38
rs906102373 Chr.13: 113821038-113821038 on Build GRCh38
rs758890844 Chr.13: 113820990-113820990 on Build GRCh38
rs746689718 Chr.13: 113820873-113820873 on Build GRCh38
rs764482337 Chr.13: 113820904-113820904 on Build GRCh38
rs778408373 Chr.13: 113820856-113820856 on Build GRCh38
rs751197313 Chr.13: 113820923-113820923 on Build GRCh38
rs41307844 Chr.13: 113820887-113820887 on Build GRCh38
rs562381726 Chr.13: 113820911-113820911 on Build GRCh38
rs1011737628 Chr.13: 113821030-113821030 on Build GRCh38

View Full Product Details