Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 77250421-77250660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALR1
Entrez Gene ID
2587
Gene Name
galanin receptor 1
Gene Aliases
GALNR, GALNR1
UniGene
Hs.272191
Gene Chromosome Location
Chr.18: 77250052-77270142 on Build GRCh38
Transcripts
NM_001480.3 XM_017025691.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1721231 c.70G>A p.G24R Chr.18: 77250618-77250618 on Build GRCh38
1389814 c.102C>T p.F34F Chr.18: 77250650-77250650 on Build GRCh38
6244809 c.103G>T p.V35F Chr.18: 77250651-77250651 on Build GRCh38
4928501 c.91G>A p.V31M Chr.18: 77250639-77250639 on Build GRCh38
4611425 c.52delC p.A20fs*23 Chr.18: 77250600-77250600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761678726 Chr.18: 77250590-77250590 on Build GRCh38
rs549249793 Chr.18: 77250510-77250510 on Build GRCh38
rs565505361 Chr.18: 77250620-77250620 on Build GRCh38
rs753457954 Chr.18: 77250602-77250602 on Build GRCh38
rs200473944 Chr.18: 77250607-77250607 on Build GRCh38
rs997549579 Chr.18: 77250481-77250481 on Build GRCh38
rs781764050 Chr.18: 77250609-77250609 on Build GRCh38
rs759188587 Chr.18: 77250606-77250606 on Build GRCh38
rs13306375 Chr.18: 77250491-77250491 on Build GRCh38
rs768301664 Chr.18: 77250583-77250583 on Build GRCh38
rs988021625 Chr.18: 77250462-77250462 on Build GRCh38
rs768873733 Chr.18: 77250626-77250626 on Build GRCh38
rs537708873 Chr.18: 77250509-77250509 on Build GRCh38
rs376635516 Chr.18: 77250493-77250493 on Build GRCh38
rs948116560 Chr.18: 77250543-77250543 on Build GRCh38
rs757711998 Chr.18: 77250608-77250608 on Build GRCh38
rs973438118 Chr.18: 77250542-77250542 on Build GRCh38
rs1031177816 Chr.18: 77250640-77250640 on Build GRCh38
rs778585624 Chr.18: 77250627-77250627 on Build GRCh38
rs968238038 Chr.18: 77250508-77250508 on Build GRCh38
rs750585637 Chr.18: 77250614-77250614 on Build GRCh38
rs546436509 Chr.18: 77250471-77250471 on Build GRCh38
rs201461949 Chr.18: 77250570-77250570 on Build GRCh38
rs1045154905 Chr.18: 77250561-77250561 on Build GRCh38
rs533177283 Chr.18: 77250444-77250444 on Build GRCh38
rs745879242 Chr.18: 77250549-77250549 on Build GRCh38
rs1002356091 Chr.18: 77250503-77250503 on Build GRCh38
rs535428789 Chr.18: 77250494-77250494 on Build GRCh38
rs780459288 Chr.18: 77250624-77250624 on Build GRCh38
rs760167095 Chr.18: 77250597-77250597 on Build GRCh38
rs749141409 Chr.18: 77250579-77250579 on Build GRCh38
rs917938039 Chr.18: 77250541-77250541 on Build GRCh38
rs752037909 Chr.18: 77250512-77250512 on Build GRCh38
rs764498666 Chr.18: 77250606-77250606 on Build GRCh38
rs1035133106 Chr.18: 77250507-77250507 on Build GRCh38
rs377602612 Chr.18: 77250552-77250552 on Build GRCh38
rs1030003729 Chr.18: 77250484-77250484 on Build GRCh38
rs755295011 Chr.18: 77250584-77250584 on Build GRCh38
rs929449672 Chr.18: 77250564-77250564 on Build GRCh38
rs577285257 Chr.18: 77250595-77250595 on Build GRCh38
rs780407897 Chr.18: 77250578-77250578 on Build GRCh38
rs375230135 Chr.18: 77250519-77250519 on Build GRCh38
rs142660460 Chr.18: 77250518-77250518 on Build GRCh38
rs942050681 Chr.18: 77250531-77250531 on Build GRCh38
rs1143093 Chr.18: 77250593-77250593 on Build GRCh38
rs12955673 Chr.18: 77250486-77250486 on Build GRCh38
rs539654659 Chr.18: 77250600-77250600 on Build GRCh38
rs758903262 Chr.18: 77250599-77250600 on Build GRCh38
rs1011286857 Chr.18: 77250478-77250478 on Build GRCh38
rs1032081992 Chr.18: 77250605-77250605 on Build GRCh38
rs771968617 Chr.18: 77250638-77250638 on Build GRCh38
rs992535113 Chr.18: 77250530-77250530 on Build GRCh38
rs1018438073 Chr.18: 77250498-77250498 on Build GRCh38

View Full Product Details