Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161723448-161723679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRLB
Entrez Gene ID
127943
Gene Name
Fc receptor like B
Gene Aliases
FCRL2, FCRLM2, FCRLY, FREB-2, FcRY
UniGene
Hs.517422
Gene Chromosome Location
Chr.1: 161721544-161728143 on Build GRCh38
Transcripts
NM_001002901.3 NM_001288829.1 NM_001288830.1 NM_001288831.1 NM_001288832.1 NM_001320241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5657280 c.166C>T p.P56S Chr.1: 161723480-161723480 on Build GRCh38
6880944 c.244G>A p.V82M Chr.1: 161723558-161723558 on Build GRCh38
4024455 c.274G>A p.V92I Chr.1: 161723588-161723588 on Build GRCh38
344873 c.262C>A p.R88R Chr.1: 161723576-161723576 on Build GRCh38
400413 c.228G>C p.E76D Chr.1: 161723542-161723542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761365982 Chr.1: 161723642-161723642 on Build GRCh38
rs201479109 Chr.1: 161723576-161723576 on Build GRCh38
rs371595857 Chr.1: 161723525-161723525 on Build GRCh38
rs567408157 Chr.1: 161723580-161723580 on Build GRCh38
rs765437491 Chr.1: 161723479-161723479 on Build GRCh38
rs780560351 Chr.1: 161723527-161723527 on Build GRCh38
rs538594471 Chr.1: 161723639-161723639 on Build GRCh38
rs376950725 Chr.1: 161723528-161723528 on Build GRCh38
rs147870526 Chr.1: 161723588-161723588 on Build GRCh38
rs771254130 Chr.1: 161723614-161723614 on Build GRCh38
rs752603524 Chr.1: 161723500-161723500 on Build GRCh38
rs777301346 Chr.1: 161723506-161723506 on Build GRCh38
rs756981625 Chr.1: 161723579-161723579 on Build GRCh38
rs760468438 Chr.1: 161723637-161723637 on Build GRCh38
rs751573615 Chr.1: 161723496-161723496 on Build GRCh38
rs774556960 Chr.1: 161723617-161723617 on Build GRCh38
rs553032070 Chr.1: 161723591-161723591 on Build GRCh38
rs541451917 Chr.1: 161723472-161723472 on Build GRCh38
rs558577824 Chr.1: 161723648-161723648 on Build GRCh38
rs767617577 Chr.1: 161723567-161723567 on Build GRCh38
rs139828673 Chr.1: 161723490-161723490 on Build GRCh38
rs150995653 Chr.1: 161723550-161723550 on Build GRCh38
rs139092332 Chr.1: 161723480-161723480 on Build GRCh38
rs367719903 Chr.1: 161723497-161723497 on Build GRCh38
rs369117765 Chr.1: 161723634-161723634 on Build GRCh38
rs759544448 Chr.1: 161723565-161723565 on Build GRCh38
rs572771679 Chr.1: 161723638-161723638 on Build GRCh38
rs139259107 Chr.1: 161723536-161723536 on Build GRCh38
rs752480579 Chr.1: 161723572-161723572 on Build GRCh38
rs762946253 Chr.1: 161723483-161723483 on Build GRCh38
rs769992593 Chr.1: 161723546-161723546 on Build GRCh38
rs544994907 Chr.1: 161723587-161723587 on Build GRCh38
rs753454454 Chr.1: 161723577-161723577 on Build GRCh38
rs538056349 Chr.1: 161723538-161723538 on Build GRCh38
rs760387437 Chr.1: 161723575-161723575 on Build GRCh38
rs759699732 Chr.1: 161723619-161723619 on Build GRCh38
rs373232160 Chr.1: 161723644-161723644 on Build GRCh38
rs767499642 Chr.1: 161723650-161723650 on Build GRCh38
rs755775561 Chr.1: 161723501-161723501 on Build GRCh38
rs778253212 Chr.1: 161723594-161723594 on Build GRCh38
rs763043596 Chr.1: 161723551-161723551 on Build GRCh38
rs769190006 Chr.1: 161723534-161723534 on Build GRCh38
rs1043618584 Chr.1: 161723606-161723606 on Build GRCh38
rs770897621 Chr.1: 161723558-161723558 on Build GRCh38
rs749443190 Chr.1: 161723596-161723596 on Build GRCh38
rs749491641 Chr.1: 161723647-161723647 on Build GRCh38
rs749995550 Chr.1: 161723585-161723585 on Build GRCh38
rs575492581 Chr.1: 161723653-161723653 on Build GRCh38
rs146761628 Chr.1: 161723517-161723517 on Build GRCh38
rs775595991 Chr.1: 161723635-161723635 on Build GRCh38
rs544043340 Chr.1: 161723656-161723656 on Build GRCh38

View Full Product Details