Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 79765098-79765367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEY1
Entrez Gene ID
23462
Gene Name
hes related family bHLH transcription factor with YRPW motif 1
Gene Aliases
BHLHb31, CHF2, HERP2, HESR1, HRT-1, OAF1, hHRT1
UniGene
Hs.234434
Gene Chromosome Location
Chr.8: 79764010-79767863 on Build GRCh38
Transcripts
NM_001040708.1 NM_001282851.1 NM_012258.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7325699 c.852delC p.T285fs*>20 Chr.8: 79765251-79765251 on Build GRCh38
7371053 c.881A>G p.Y294C Chr.8: 79765222-79765222 on Build GRCh38
6583484 c.779C>A p.S260Y Chr.8: 79765336-79765336 on Build GRCh38
3835050 c.888T>A p.P296P Chr.8: 79765215-79765215 on Build GRCh38
3835052 c.837G>A p.L279L Chr.8: 79765266-79765266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567381029 Chr.8: 79765169-79765169 on Build GRCh38
rs571606795 Chr.8: 79765160-79765160 on Build GRCh38
rs3175191 Chr.8: 79765348-79765348 on Build GRCh38
rs150402060 Chr.8: 79765200-79765200 on Build GRCh38
rs769834959 Chr.8: 79765346-79765346 on Build GRCh38
rs919305445 Chr.8: 79765207-79765207 on Build GRCh38
rs755216317 Chr.8: 79765328-79765328 on Build GRCh38
rs933123959 Chr.8: 79765287-79765287 on Build GRCh38
rs934924996 Chr.8: 79765195-79765195 on Build GRCh38
rs774080653 Chr.8: 79765241-79765241 on Build GRCh38
rs769255917 Chr.8: 79765314-79765314 on Build GRCh38
rs112785069 Chr.8: 79765350-79765350 on Build GRCh38
rs1042490959 Chr.8: 79765290-79765290 on Build GRCh38
rs534875773 Chr.8: 79765140-79765140 on Build GRCh38
rs781040574 Chr.8: 79765333-79765333 on Build GRCh38
rs950253047 Chr.8: 79765242-79765242 on Build GRCh38
rs145511397 Chr.8: 79765249-79765249 on Build GRCh38
rs1013015043 Chr.8: 79765135-79765135 on Build GRCh38
rs371598666 Chr.8: 79765303-79765303 on Build GRCh38
rs765771783 Chr.8: 79765298-79765298 on Build GRCh38
rs917422406 Chr.8: 79765224-79765224 on Build GRCh38
rs971484911 Chr.8: 79765155-79765155 on Build GRCh38
rs139456432 Chr.8: 79765145-79765145 on Build GRCh38
rs978862126 Chr.8: 79765147-79765147 on Build GRCh38
rs1026271805 Chr.8: 79765344-79765344 on Build GRCh38
rs547520315 Chr.8: 79765320-79765320 on Build GRCh38
rs770437939 Chr.8: 79765240-79765240 on Build GRCh38
rs750455935 Chr.8: 79765299-79765299 on Build GRCh38
rs747999619 Chr.8: 79765345-79765345 on Build GRCh38
rs748087859 Chr.8: 79765187-79765187 on Build GRCh38
rs145654005 Chr.8: 79765212-79765212 on Build GRCh38
rs925611598 Chr.8: 79765177-79765177 on Build GRCh38
rs909103181 Chr.8: 79765216-79765216 on Build GRCh38
rs368116797 Chr.8: 79765222-79765222 on Build GRCh38
rs769922531 Chr.8: 79765191-79765191 on Build GRCh38
rs754257700 Chr.8: 79765273-79765273 on Build GRCh38
rs61753715 Chr.8: 79765292-79765292 on Build GRCh38
rs1005606433 Chr.8: 79765289-79765289 on Build GRCh38
rs191861249 Chr.8: 79765215-79765215 on Build GRCh38
rs866901091 Chr.8: 79765316-79765316 on Build GRCh38
rs964437523 Chr.8: 79765144-79765144 on Build GRCh38
rs764532628 Chr.8: 79765261-79765261 on Build GRCh38

View Full Product Details